Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561678_m1
          See other NPR1 GE Assays ›
          Gene Symbol:
          Npr1
          Gene Name:
          natriuretic peptide receptor 1
          Gene Aliases:
          Anpra, Gca, NPR-A, Npra
          Chromosome Location:
          Chr.2: 189840403 - 189857034 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561678_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Npr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012613.1 NP_036745.1 5 - 6 1618 88
          XM_006232591.2 XP_006232653.1 6 - 7 2119 88
          GenBank mRNA BC128742.1 - 5 - 6 1604 7458324 88
          M74535.1 - 6 - 7 1567 88
          X14773.1 - 5 - 6 1475 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004102.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Npr1

          Panther Classification:

          Molecular Function -

          guanylate cyclase

          Gene Ontology Categories:

          Function(s) Process(es)

          cGMP biosynthetic process
          protein phosphorylation
          cell surface receptor signaling pathway
          receptor guanylyl cyclase signaling pathway
          regulation of blood pressure
          positive regulation of cGMP biosynthetic process
          intracellular signal transduction
          dopamine metabolic process
          negative regulation of smooth muscle cell proliferation
          regulation of blood vessel size
          guanylate cyclase activity
          protein kinase activity
          protein binding
          ATP binding
          GTP binding
          natriuretic peptide receptor activity
          peptide hormone binding
          protein kinase binding
          hormone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52983 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15245 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15245 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15245 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15245 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75844 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71306 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8657 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10824661 - -
          Affymetrix Rat Gene 1.0 ST Array 10824656 - -
          Affymetrix Rat Gene 1.0 ST Array 10824659 - -
          Affymetrix Rat Gene 1.0 ST Array 10824673 - -
          Affymetrix Rat Gene 1.0 ST Array 10824665 - -
          Affymetrix Rat Gene 1.0 ST Array 10824677 - -
          Affymetrix Rat Gene 1.0 ST Array 10824679 - -
          Affymetrix Rat Gene 1.0 ST Array 10824662 - -
          Affymetrix Rat Gene 1.0 ST Array 10824668 - -
          Affymetrix Rat Gene 1.0 ST Array 10824676 - -
          Affymetrix Rat Gene 1.0 ST Array 10824669 - -
          Affymetrix Rat Gene 1.0 ST Array 10824671 - -
          Affymetrix Rat Gene 1.0 ST Array 10824672 - -
          Affymetrix Rat Gene 1.0 ST Array 10824678 - -
          Affymetrix Rat Gene 1.0 ST Array 10824657 - -
          Affymetrix Rat Gene 1.0 ST Array 10824674 - -
          Affymetrix Rat Gene 1.0 ST Array 10824667 - -
          Affymetrix Rat Gene 1.0 ST Array 10824663 - -
          Affymetrix Rat Gene 1.0 ST Array 10824664 - -
          Affymetrix Rat Gene 1.0 ST Array 10824666 - -
          Affymetrix Rat Gene 1.0 ST Array 10824670 - -
          Affymetrix Rat Gene 1.0 ST Array 10824660 - -
          Affymetrix Rat Gene 1.0 ST Array 10824658 - -
          Affymetrix Rat Gene 1.0 ST Array 10824675 - -
          Affymetrix Rat Gene 1.1 ST Array 10824655 - -
          Rat Genome 230 2.0 Array 1368201_at - -
          Rat Genome 230 2.0 Array 1368201_PM_at - -
          Affymetrix HT Rat Focus Array 1368201_at - -
          Affymetrix HT Rat Focus Array 1368201_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200028052.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028054.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028048.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028051.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028039.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028047.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028045.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028050.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028058.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028059.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028035.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028037.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028036.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028038.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028040.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028041.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028042.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028044.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028046.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028053.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028043.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028049.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028055.rn.1 TC0200004102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028056.rn.1 TC0200004102.rn.1 RTA TC
          Rat Expression Set 230 1368201_at - -
          Rat Expression Set 230 1368201_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10824655 - -
          Rat Genome U34 Set S80456_s_at - -
          Rat Genome U34 Set S82229_s_at - -
          Rat Genome U34 Set J05677mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21801185 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360992 - -
          RatRef-12 Expression BeadChip ILMN_58902 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline