Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562086_m1
          See other TSC2 GE Assays ›
          Gene Symbol:
          Tsc2
          Gene Name:
          tuberous sclerosis 2
          Gene Aliases:
          Rc
          Chromosome Location:
          Chr.10: 13962005 - 13996751 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562086_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012680.3 NP_036812.2 14 - 15 1525 66
          XM_006245909.2 XP_006245971.1 14 - 15 1706 66
          XM_006245910.2 XP_006245972.1 14 - 15 1706 66
          XM_006245911.2 XP_006245973.1 14 - 15 1706 66
          XM_006245912.2 XP_006245974.1 14 - 15 1703 66
          XM_006245913.2 XP_006245975.1 14 - 15 1706 66
          XM_006245914.2 XP_006245976.1 14 - 15 1706 66
          XM_006245915.2 XP_006245977.1 14 - 15 1706 66
          XM_006245916.2 XP_006245978.1 14 - 15 1706 66
          XM_006245917.2 XP_006245979.1 14 - 15 1703 66
          XM_008767543.1 XP_008765765.1 14 - 15 1703 66
          XM_008767544.1 XP_008765766.1 13 - 14 1485 66
          GenBank mRNA D50413.1 - 13 - 14 1477 66
          U24150.1 - 14 - 15 1525 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002393.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsc2

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          neural tube closure
          negative regulation of protein kinase activity
          protein import into nucleus
          acute-phase response
          signal transduction
          heart development
          protein localization
          negative regulation of cell proliferation
          phosphatidylinositol 3-kinase signaling
          negative regulation of phosphatidylinositol 3-kinase signaling
          cell projection organization
          regulation of endocytosis
          negative regulation of Wnt signaling pathway
          negative regulation of TOR signaling
          cellular response to insulin stimulus
          negative regulation of MAP kinase activity
          protein kinase B signaling
          positive regulation of GTPase activity
          protein transport into plasma membrane raft
          establishment of protein localization
          negative regulation of cell size
          positive regulation of transcription from RNA polymerase II promoter
          glucose import
          regulation of insulin receptor signaling pathway
          negative regulation of insulin receptor signaling pathway
          insulin-like growth factor receptor signaling pathway
          negative regulation of epithelial cell proliferation
          positive chemotaxis
          regulation of small GTPase mediated signal transduction
          protein homooligomerization
          protein heterooligomerization
          regulation of cell cycle
          negative regulation of protein kinase B signaling
          GTPase activator activity
          protein binding
          phosphatase binding
          protein homodimerization activity
          protein heterodimerization activity
          14-3-3 protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P148338 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P024252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111779 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178182 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166169 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P024252 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148338 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148338 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P024252 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P024252 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P148338 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P471578 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700514111 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10557 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66683 - -
          Agilent Rat cDNA Microarray (G4105A) 700514111 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741150 - -
          Affymetrix Rat Gene 1.0 ST Array 10741175 - -
          Affymetrix Rat Gene 1.0 ST Array 10741183 - -
          Affymetrix Rat Gene 1.0 ST Array 10741155 - -
          Affymetrix Rat Gene 1.0 ST Array 10741152 - -
          Affymetrix Rat Gene 1.0 ST Array 10741178 - -
          Affymetrix Rat Gene 1.0 ST Array 10741167 - -
          Affymetrix Rat Gene 1.0 ST Array 10741161 - -
          Affymetrix Rat Gene 1.0 ST Array 10741181 - -
          Affymetrix Rat Gene 1.0 ST Array 10741179 - -
          Affymetrix Rat Gene 1.0 ST Array 10741186 - -
          Affymetrix Rat Gene 1.0 ST Array 10741144 - -
          Affymetrix Rat Gene 1.0 ST Array 10741160 - -
          Affymetrix Rat Gene 1.0 ST Array 10741165 - -
          Affymetrix Rat Gene 1.0 ST Array 10741157 - -
          Affymetrix Rat Gene 1.0 ST Array 10741166 - -
          Affymetrix Rat Gene 1.0 ST Array 10741151 - -
          Affymetrix Rat Gene 1.0 ST Array 10741188 - -
          Affymetrix Rat Gene 1.0 ST Array 10741171 - -
          Affymetrix Rat Gene 1.0 ST Array 10741176 - -
          Affymetrix Rat Gene 1.0 ST Array 10741158 - -
          Affymetrix Rat Gene 1.0 ST Array 10741177 - -
          Affymetrix Rat Gene 1.0 ST Array 10741180 - -
          Affymetrix Rat Gene 1.0 ST Array 10741164 - -
          Affymetrix Rat Gene 1.0 ST Array 10741159 - -
          Affymetrix Rat Gene 1.0 ST Array 10741170 - -
          Affymetrix Rat Gene 1.0 ST Array 10741173 - -
          Affymetrix Rat Gene 1.0 ST Array 10741184 - -
          Affymetrix Rat Gene 1.0 ST Array 10741146 - -
          Affymetrix Rat Gene 1.0 ST Array 10741154 - -
          Affymetrix Rat Gene 1.0 ST Array 10741168 - -
          Affymetrix Rat Gene 1.0 ST Array 10741172 - -
          Affymetrix Rat Gene 1.0 ST Array 10741145 - -
          Affymetrix Rat Gene 1.0 ST Array 10741187 - -
          Affymetrix Rat Gene 1.0 ST Array 10741153 - -
          Affymetrix Rat Gene 1.0 ST Array 10741148 - -
          Affymetrix Rat Gene 1.0 ST Array 10741156 - -
          Affymetrix Rat Gene 1.0 ST Array 10741174 - -
          Affymetrix Rat Gene 1.0 ST Array 10741163 - -
          Affymetrix Rat Gene 1.0 ST Array 10741162 - -
          Affymetrix Rat Gene 1.0 ST Array 10741182 - -
          Affymetrix Rat Gene 1.0 ST Array 10741185 - -
          Affymetrix Rat Gene 1.0 ST Array 10741149 - -
          Affymetrix Rat Gene 1.0 ST Array 10741169 - -
          Affymetrix Rat Gene 1.1 ST Array 10741143 - -
          Rat Genome 230 2.0 Array 1368056_at - -
          Rat Genome 230 2.0 Array 1368056_PM_at - -
          Affymetrix HT Rat Focus Array 1368056_PM_at - -
          Affymetrix HT Rat Focus Array 1368056_at - -
          Affymetrix Rat Clariom S Assay PSR1000021338.rn.1 TC1000002393.rn.1 RTA TC
          Rat Expression Set 230 1368056_at - -
          Rat Expression Set 230 1368056_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741143 - -
          Rat Genome U34 Set U24150_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373092 - -
          RatRef-12 Expression BeadChip ILMN_57872 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0