Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562500_m1
          See other TH GE Assays ›
          Gene Symbol:
          Th
          Gene Name:
          tyrosine hydroxylase
          Gene Aliases:
          The
          Chromosome Location:
          Chr.1: 216073034 - 216080287 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562500_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Th
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012740.3 NP_036872.1 12 - 13 1346 60
          GenBank mRNA M10244.1 - 12 - 13 1346 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006990.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Th

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          synaptic transmission, dopaminergic
          response to amphetamine
          dopamine biosynthetic process from tyrosine
          fatty acid metabolic process
          sphingolipid metabolic process
          heart development
          visual perception
          sensory perception of sound
          learning
          memory
          mating behavior
          locomotory behavior
          regulation of heart contraction
          response to water deprivation
          response to light stimulus
          response to herbicide
          response to salt stress
          organ morphogenesis
          response to metal ion
          response to zinc ion
          multicellular organism aging
          response to organic cyclic compound
          response to activity
          aminergic neurotransmitter loading into synaptic vesicle
          glycoside metabolic process
          response to insecticide
          phthalate metabolic process
          cerebral cortex development
          response to nutrient levels
          response to estradiol
          response to lipopolysaccharide
          isoquinoline alkaloid metabolic process
          response to nicotine
          social behavior
          cellular response to drug
          response to isolation stress
          response to immobilization stress
          neurotransmitter biosynthetic process
          terpene metabolic process
          dopamine biosynthetic process
          epinephrine biosynthetic process
          norepinephrine biosynthetic process
          catecholamine biosynthetic process
          eye photoreceptor cell development
          response to drug
          circadian sleep/wake cycle
          eating behavior
          response to peptide hormone
          response to ethanol
          response to ether
          response to pyrethroid
          response to steroid hormone
          embryonic camera-type eye morphogenesis
          cognition
          protein homotetramerization
          response to corticosterone
          response to electrical stimulus
          phytoalexin metabolic process
          oxidation-reduction process
          response to growth factor
          cellular response to manganese ion
          cellular response to alkaloid
          cellular response to nicotine
          cellular response to glucose stimulus
          cellular response to growth factor stimulus
          monooxygenase activity
          tyrosine 3-monooxygenase activity
          protein binding
          ferrous iron binding
          ferric iron binding
          amino acid binding
          oxygen binding
          enzyme binding
          protein domain specific binding
          tetrahydrobiopterin binding
          dopamine binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P039656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744905 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14186 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039656 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039656 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P039656 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P301729 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700183273 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35578 - -
          Agilent Rat cDNA Microarray (G4105A) 700183273 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727013 - -
          Affymetrix Rat Gene 1.0 ST Array 10727023 - -
          Affymetrix Rat Gene 1.0 ST Array 10727016 - -
          Affymetrix Rat Gene 1.0 ST Array 10727017 - -
          Affymetrix Rat Gene 1.0 ST Array 10727018 - -
          Affymetrix Rat Gene 1.0 ST Array 10727022 - -
          Affymetrix Rat Gene 1.0 ST Array 10727012 - -
          Affymetrix Rat Gene 1.0 ST Array 10727020 - -
          Affymetrix Rat Gene 1.0 ST Array 10727021 - -
          Affymetrix Rat Gene 1.0 ST Array 10727019 - -
          Affymetrix Rat Gene 1.0 ST Array 10727015 - -
          Affymetrix Rat Gene 1.0 ST Array 10727024 - -
          Affymetrix Rat Gene 1.0 ST Array 10727014 - -
          Affymetrix Rat Gene 1.1 ST Array 10727011 - -
          Rat Genome 230 2.0 Array 1373443_PM_a_at - -
          Rat Genome 230 2.0 Array 1373443_a_at - -
          Rat Genome 230 2.0 Array 1387075_at - -
          Rat Genome 230 2.0 Array 1387075_PM_at - -
          Affymetrix HT Rat Focus Array 1387075_at - -
          Affymetrix HT Rat Focus Array 1387075_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100055275.rn.1 TC0100006990.rn.1 RTA TC
          Rat Expression Set 230 1387075_at - -
          Rat Expression Set 230 1387075_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10727011 - -
          Rat Genome U34 Set M10244_at - -
          Rat Genome U34 Set M10244_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20728591 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360569 - -
          RatRef-12 Expression BeadChip ILMN_58975 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline