Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562794_m1
          See other LYZ2 GE Assays ›
          Gene Symbol:
          Lyz2
          Gene Name:
          lysozyme 2
          Gene Aliases:
          Lysz, Lyz, Lyz1
          Chromosome Location:
          Chr.7: 60335968 - 60341264 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562794_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lyz2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012771.3 NP_036903.2 1 - 2 165 92
          GenBank mRNA BC058490.1 - 1 - 2 169 6919643 92
          FQ221278.1 - 1 - 2 165 92
          FQ226475.1 - 1 - 2 165 92
          FQ226588.1 - 1 - 2 165 92
          FQ227076.1 - 1 - 2 165 92
          FQ227452.1 - 1 - 2 165 92
          FQ227932.1 - 1 - 2 165 92
          FQ228036.1 - 1 - 2 165 92
          FQ229300.1 - 1 - 2 165 92
          FQ230656.1 - 1 - 2 165 92
          FQ231085.1 - 1 - 2 165 92
          FQ231872.1 - 1 - 2 165 92
          FQ231961.1 - 1 - 2 92
          FQ233423.1 - 1 - 2 165 92
          FQ233793.1 - 1 - 2 165 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002754.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lyz2

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          metabolic process
          cytolysis
          defense response to bacterium
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          lysozyme activity
          hydrolase activity, acting on glycosyl bonds
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P025286 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025286 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025286 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P025286 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10020 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86415 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10902549 - -
          Affymetrix Rat Gene 1.0 ST Array 10902551 - -
          Affymetrix Rat Gene 1.0 ST Array 10895588 - -
          Affymetrix Rat Gene 1.0 ST Array 10902548 - -
          Affymetrix Rat Gene 1.0 ST Array 10902550 - -
          Affymetrix Rat Gene 1.1 ST Array 10895585 - -
          Affymetrix Rat Gene 1.1 ST Array 10902547 - -
          Rat Genome 230 2.0 Array 1370154_PM_at - -
          Rat Genome 230 2.0 Array 1370154_at - -
          Affymetrix HT Rat Focus Array 1370154_PM_at - -
          Affymetrix HT Rat Focus Array 1370154_at - -
          Affymetrix Rat Clariom S Assay PSR0700021438.rn.1 TC0700002754.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021440.rn.1 TC0700002754.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021446.rn.1 TC0700002754.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021447.rn.1 TC0700002754.rn.1 RTA TC
          Rat Expression Set 230 1370154_at - -
          Rat Expression Set 230 1370154_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10902547 - -
          Rat Genome U34 Set rc_AA892775_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22178267 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline