Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564116_m1
          See other FGF9 GE Assays ›
          Gene Symbol:
          Fgf9
          Gene Name:
          fibroblast growth factor 9
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 38341657 - 38386945 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564116_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgf9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012952.1 NP_037084.1 1 - 2 437 75
          GenBank mRNA BC078699.1 - 1040 75
          D14839.1 - 1 - 2 456 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000581.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fgf9

          Panther Classification:

          Molecular Function -

          growth factor intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          osteoblast differentiation
          positive regulation of mesenchymal cell proliferation
          chondrocyte differentiation
          protein import into nucleus
          cell-cell signaling
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          male gonad development
          positive regulation of gene expression
          substantia nigra development
          negative regulation of Wnt signaling pathway
          male sex determination
          lung development
          embryonic limb morphogenesis
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          positive regulation of activin receptor signaling pathway
          inner ear morphogenesis
          positive regulation of MAPK cascade
          positive regulation of smoothened signaling pathway
          regulation of timing of cell differentiation
          embryonic digestive tract development
          embryonic skeletal system development
          positive regulation of epithelial cell proliferation
          positive regulation of cell division
          positive regulation of cardiac muscle cell proliferation
          lung-associated mesenchyme development
          positive regulation of canonical Wnt signaling pathway
          fibroblast growth factor receptor binding
          growth factor activity
          heparin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P486914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73571 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144038 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72908 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12184 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110684 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P486914 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P486914 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P486914 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P486914 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70546 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59349 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49355 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72571 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10780814 - -
          Affymetrix Rat Gene 1.0 ST Array 10780816 - -
          Affymetrix Rat Gene 1.0 ST Array 10780818 - -
          Affymetrix Rat Gene 1.0 ST Array 10780817 - -
          Affymetrix Rat Gene 1.0 ST Array 10780815 - -
          Affymetrix Rat Gene 1.1 ST Array 10780813 - -
          Rat Genome 230 2.0 Array 1392865_PM_at - -
          Rat Genome 230 2.0 Array 1370747_at - -
          Rat Genome 230 2.0 Array 1370747_PM_at - -
          Rat Genome 230 2.0 Array 1392865_at - -
          Affymetrix HT Rat Focus Array 1392865_PM_at - -
          Affymetrix HT Rat Focus Array 1370747_PM_at - -
          Affymetrix HT Rat Focus Array 1392865_at - -
          Affymetrix HT Rat Focus Array 1370747_at - -
          Affymetrix Rat Clariom S Assay PSR1500004081.rn.1 TC1500000581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500004077.rn.1 TC1500000581.rn.1 RTA TC
          Rat Expression Set 230 1370747_PM_at - -
          Rat Expression Set 230 1370747_at - -
          Rat Expression Set 230 1392865_at - -
          Rat Expression Set 230 1392865_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10780813 - -
          Rat Genome U34 Set D14839_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21518483 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365421 - -
          RatRef-12 Expression BeadChip ILMN_53791 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline