Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565242_m1
          See other SDC2 GE Assays ›
          Gene Symbol:
          Sdc2
          Gene Name:
          syndecan 2
          Gene Aliases:
          HSPG
          Chromosome Location:
          Chr.7: 71572731 - 71686139 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565242_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sdc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013082.3 NP_037214.1 3 - 4 690 82
          XM_006241518.2 XP_006241580.1 3 - 4 902 82
          GenBank mRNA BC070890.1 - 3 - 4 480 7098631 82
          FQ218442.1 - 3 - 4 423 82
          FQ219283.1 - 3 - 4 724 82
          FQ219510.1 - 3 - 4 722 82
          FQ222974.1 - 3 - 4 772 82
          M81687.1 - 3 - 4 690 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000925.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sdc2

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          response to caffeine
          wound healing
          dendrite morphogenesis
          regulation of dendrite morphogenesis
          protein binding
          PDZ domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11701 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675116 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152794 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128389 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114167 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31297 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138005 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27758 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124411 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163382 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140262 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167072 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30296 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11701 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11701 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11701 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11193 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11193 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11701 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700610226 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55358 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59027 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45626 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6705 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56097 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8709 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80108 - -
          Agilent Rat cDNA Microarray (G4105A) 700610226 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10896027 - -
          Affymetrix Rat Gene 1.0 ST Array 10896023 - -
          Affymetrix Rat Gene 1.0 ST Array 10896025 - -
          Affymetrix Rat Gene 1.0 ST Array 10896021 - -
          Affymetrix Rat Gene 1.0 ST Array 10896024 - -
          Affymetrix Rat Gene 1.0 ST Array 10896022 - -
          Affymetrix Rat Gene 1.0 ST Array 10896026 - -
          Affymetrix Rat Gene 1.1 ST Array 10896020 - -
          Rat Genome 230 2.0 Array 1370166_at - -
          Rat Genome 230 2.0 Array 1370167_at - -
          Rat Genome 230 2.0 Array 1370166_PM_at - -
          Rat Genome 230 2.0 Array 1382189_at - -
          Rat Genome 230 2.0 Array 1382189_PM_at - -
          Rat Genome 230 2.0 Array 1370167_PM_at - -
          Affymetrix HT Rat Focus Array 1370167_PM_at - -
          Affymetrix HT Rat Focus Array 1370166_at - -
          Affymetrix HT Rat Focus Array 1370167_at - -
          Affymetrix HT Rat Focus Array 1382189_PM_at - -
          Affymetrix HT Rat Focus Array 1382189_at - -
          Affymetrix HT Rat Focus Array 1370166_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700007069.rn.1 TC0700000925.rn.1 RTA TC
          Rat Expression Set 230 1370167_PM_at - -
          Rat Expression Set 230 1370166_PM_at - -
          Rat Expression Set 230 1370167_at - -
          Rat Expression Set 230 1370166_at - -
          Rat Expression Set 230 1382189_at - -
          Rat Expression Set 230 1382189_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10896020 - -
          Rat Genome U34 Set M81687_at - -
          Rat Genome U34 Set rc_AI011678_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20707257 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361950 - -
          RatRef-12 Expression BeadChip ILMN_61221 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline