Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566466_m1
          See other C3 GE Assays ›
          Gene Symbol:
          C3
          Gene Name:
          complement component 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 9721137 - 9747084 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566466_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016994.2 NP_058690.2 27 - 28 3532 72
          GenBank mRNA CO796620.1 - 4 - 5 399 72
          X52477.1 - 27 - 28 3532 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000078.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C3

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of type IIa hypersensitivity
          positive regulation of protein phosphorylation
          positive regulation of activation of membrane attack complex
          tolerance induction
          fatty acid metabolic process
          chemotaxis
          inflammatory response
          complement activation
          complement activation, alternative pathway
          complement activation, classical pathway
          blood coagulation
          positive regulation of vascular endothelial growth factor production
          positive regulation of glucose transport
          regulation of triglyceride biosynthetic process
          positive regulation of lipid storage
          negative regulation of endopeptidase activity
          response to magnesium ion
          response to estradiol
          response to progesterone
          response to estrogen
          positive regulation of G-protein coupled receptor protein signaling pathway
          positive regulation of angiogenesis
          positive regulation of developmental growth
          positive regulation of phagocytosis
          response to glucocorticoid
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of apoptotic cell clearance
          endopeptidase inhibitor activity
          protein binding
          lipid binding
          C5L2 anaphylatoxin chemotactic receptor binding
          cofactor binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1023538 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1023538 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1023538 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1023538 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1023538 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12794 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36781 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67099 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52266 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20805 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10931752 - -
          Affymetrix Rat Gene 1.0 ST Array 10931729 - -
          Affymetrix Rat Gene 1.0 ST Array 10931737 - -
          Affymetrix Rat Gene 1.0 ST Array 10931736 - -
          Affymetrix Rat Gene 1.0 ST Array 10931758 - -
          Affymetrix Rat Gene 1.0 ST Array 10931747 - -
          Affymetrix Rat Gene 1.0 ST Array 10931744 - -
          Affymetrix Rat Gene 1.0 ST Array 10931755 - -
          Affymetrix Rat Gene 1.0 ST Array 10931741 - -
          Affymetrix Rat Gene 1.0 ST Array 10931750 - -
          Affymetrix Rat Gene 1.0 ST Array 10931749 - -
          Affymetrix Rat Gene 1.0 ST Array 10931738 - -
          Affymetrix Rat Gene 1.0 ST Array 10931733 - -
          Affymetrix Rat Gene 1.0 ST Array 10931743 - -
          Affymetrix Rat Gene 1.0 ST Array 10931754 - -
          Affymetrix Rat Gene 1.0 ST Array 10931728 - -
          Affymetrix Rat Gene 1.0 ST Array 10931725 - -
          Affymetrix Rat Gene 1.0 ST Array 10931757 - -
          Affymetrix Rat Gene 1.0 ST Array 10931746 - -
          Affymetrix Rat Gene 1.0 ST Array 10931734 - -
          Affymetrix Rat Gene 1.0 ST Array 10931727 - -
          Affymetrix Rat Gene 1.0 ST Array 10931723 - -
          Affymetrix Rat Gene 1.0 ST Array 10931722 - -
          Affymetrix Rat Gene 1.0 ST Array 10931720 - -
          Affymetrix Rat Gene 1.0 ST Array 10931753 - -
          Affymetrix Rat Gene 1.0 ST Array 10931732 - -
          Affymetrix Rat Gene 1.0 ST Array 10931742 - -
          Affymetrix Rat Gene 1.0 ST Array 10931735 - -
          Affymetrix Rat Gene 1.0 ST Array 10931740 - -
          Affymetrix Rat Gene 1.0 ST Array 10931726 - -
          Affymetrix Rat Gene 1.0 ST Array 10931739 - -
          Affymetrix Rat Gene 1.0 ST Array 10931731 - -
          Affymetrix Rat Gene 1.0 ST Array 10931748 - -
          Affymetrix Rat Gene 1.0 ST Array 10931718 - -
          Affymetrix Rat Gene 1.0 ST Array 10931719 - -
          Affymetrix Rat Gene 1.0 ST Array 10931756 - -
          Affymetrix Rat Gene 1.0 ST Array 10931745 - -
          Affymetrix Rat Gene 1.0 ST Array 10931751 - -
          Affymetrix Rat Gene 1.0 ST Array 10931724 - -
          Affymetrix Rat Gene 1.0 ST Array 10931721 - -
          Affymetrix Rat Gene 1.0 ST Array 10931730 - -
          Affymetrix Rat Gene 1.1 ST Array 10931717 - -
          Rat Genome 230 2.0 Array 1368000_at - -
          Rat Genome 230 2.0 Array 1368000_PM_at - -
          Affymetrix HT Rat Focus Array 1368000_PM_at - -
          Affymetrix HT Rat Focus Array 1368000_at - -
          Affymetrix Rat Clariom S Assay PSR0900000468.rn.1 TC0900000078.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900000469.rn.1 TC0900000078.rn.1 RTA TC
          Rat Expression Set 230 1368000_at - -
          Rat Expression Set 230 1368000_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10931717 - -
          Rat Genome U34 Set M29866_s_at - -
          Rat Genome U34 Set X52477_at - -
          Rat Genome U34 Set M29866_s_at - -
          Rat Genome U34 Set X52477_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22158850 - -

          Back To Top
           

          Citations (1)

          Early postnatal oestradiol exposure causes insulin resistance and signs of inflammation in circulation and skeletal muscle.
          Authors
          Alexanderson C, Eriksson E, Stener-Victorin E, Lonn M, Holmang A
          Journal
          J Endocrinol - Page No 49 (Wed Apr 01 00:00:00 UTC 2009)
          Volume (Year)
          201 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline