Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567532_m1
          See other SQLE GE Assays ›
          Gene Symbol:
          Sqle
          Gene Name:
          squalene epoxidase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 99609929 - 99624803 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567532_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sqle
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017136.2 NP_058832.1 2 - 3 610 77
          GenBank mRNA BC097330.1 - 2 - 3 610 7460860 77
          D37920.1 - 2 - 3 741 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001177.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sqle

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular aromatic compound metabolic process
          cholesterol metabolic process
          response to organic substance
          oxidation-reduction process
          squalene monooxygenase activity
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P012232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155954 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141278 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012232 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012232 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P012232 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P365580 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700491942 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19019 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79331 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49099 - -
          Agilent Rat cDNA Microarray (G4105A) 700491942 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10896782 - -
          Affymetrix Rat Gene 1.0 ST Array 10896774 - -
          Affymetrix Rat Gene 1.0 ST Array 10896784 - -
          Affymetrix Rat Gene 1.0 ST Array 10896778 - -
          Affymetrix Rat Gene 1.0 ST Array 10896779 - -
          Affymetrix Rat Gene 1.0 ST Array 10896783 - -
          Affymetrix Rat Gene 1.0 ST Array 10896776 - -
          Affymetrix Rat Gene 1.0 ST Array 10896773 - -
          Affymetrix Rat Gene 1.0 ST Array 10896775 - -
          Affymetrix Rat Gene 1.0 ST Array 10896780 - -
          Affymetrix Rat Gene 1.0 ST Array 10896781 - -
          Affymetrix Rat Gene 1.0 ST Array 10896777 - -
          Affymetrix Rat Gene 1.1 ST Array 10896772 - -
          Rat Genome 230 2.0 Array 1387017_PM_at - -
          Rat Genome 230 2.0 Array 1387017_at - -
          Affymetrix HT Rat Focus Array 1387017_PM_at - -
          Affymetrix HT Rat Focus Array 1387017_at - -
          Affymetrix Rat Clariom S Assay PSR0700008618.rn.1 TC0700001177.rn.1 RTA TC
          Rat Expression Set 230 1387017_at - -
          Rat Expression Set 230 1387017_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10896772 - -
          Rat Genome U34 Set D37920_at - -
          Rat Genome U34 Set rc_AA819300_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21385463 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364854 - -
          RatRef-12 Expression BeadChip ILMN_70256 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline