Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567668_m1
          See other ADORA1 GE Assays ›
          Gene Symbol:
          Adora1
          Gene Name:
          adenosine A1 receptor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 51042246 - 51076856 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567668_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adora1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017155.2 NP_058851.2 1 - 2 403 90
          XM_006249859.2 XP_006249921.1 2 - 3 694 90
          XM_006249860.2 XP_006249922.1 2 - 3 469 90
          XM_006249861.2 XP_006249923.1 2 - 3 633 90
          XM_008769552.1 XP_008767774.1 2 - 3 856 90
          XM_008769553.1 XP_008767775.1 2 - 3 795 90
          GenBank mRNA AB001089.1 - 1 - 2 340 90
          AF042079.1 - 1 - 2 371 90
          M69045.1 - 1 - 2 403 90
          Y12519.1 - 1 - 2 755 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001437.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adora1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          temperature homeostasis
          response to hypoxia
          adenosine receptor signaling pathway
          regulation of respiratory gaseous exchange by neurological system process
          negative regulation of acute inflammatory response
          negative regulation of leukocyte migration
          positive regulation of peptide secretion
          regulation of glomerular filtration
          protein targeting to membrane
          G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          negative regulation of cell proliferation
          negative regulation of glutamate secretion
          lipid catabolic process
          negative regulation of synaptic transmission, GABAergic
          positive regulation of nucleoside transport
          negative regulation of neurotrophin production
          positive regulation of protein dephosphorylation
          negative regulation of renal sodium excretion
          regulation of vasodilation
          negative regulation of circadian sleep/wake cycle, sleep
          negative regulation of circadian sleep/wake cycle, non-REM sleep
          negative regulation of apoptotic process
          positive regulation of potassium ion transport
          positive regulation of epidermal growth factor-activated receptor activity
          negative regulation of blood pressure
          positive regulation of blood pressure
          negative regulation of heart contraction
          negative regulation of vasodilation
          negative regulation of hormone secretion
          regulation of synaptic plasticity
          negative regulation of inflammatory response
          cognition
          detection of temperature stimulus involved in sensory perception of pain
          negative regulation of lipid catabolic process
          regulation of sensory perception of pain
          negative regulation of synaptic transmission, glutamatergic
          regulation of cardiac muscle contraction
          negative regulation of cardiac muscle contraction
          excitatory postsynaptic potential
          relaxation of vascular smooth muscle
          negative regulation of mucus secretion
          negative regulation of long term synaptic depression
          G-protein coupled adenosine receptor activity
          G-protein coupled receptor binding
          purine nucleoside binding
          phospholipase C activity
          protein binding
          G-protein beta/gamma-subunit complex binding
          heterotrimeric G-protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P137633 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P004477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909086 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81515 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64880 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88486 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P137633 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004477 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004477 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P137633 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P004477 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P137633 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P447514 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P137633 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72948 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21425 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29442 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767773 - -
          Affymetrix Rat Gene 1.0 ST Array 10767772 - -
          Affymetrix Rat Gene 1.0 ST Array 10767777 - -
          Affymetrix Rat Gene 1.0 ST Array 10767779 - -
          Affymetrix Rat Gene 1.0 ST Array 10767778 - -
          Affymetrix Rat Gene 1.0 ST Array 10767775 - -
          Affymetrix Rat Gene 1.0 ST Array 10767776 - -
          Affymetrix Rat Gene 1.0 ST Array 10767774 - -
          Affymetrix Rat Gene 1.0 ST Array 10767780 - -
          Affymetrix Rat Gene 1.1 ST Array 10767771 - -
          Rat Genome 230 2.0 Array 1370584_PM_a_at - -
          Rat Genome 230 2.0 Array 1370584_a_at - -
          Rat Genome 230 2.0 Array 1387721_at - -
          Rat Genome 230 2.0 Array 1387721_PM_at - -
          Affymetrix HT Rat Focus Array 1387721_at - -
          Affymetrix HT Rat Focus Array 1370584_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387721_PM_at - -
          Affymetrix HT Rat Focus Array 1370584_a_at - -
          Affymetrix Rat Clariom S Assay PSR1300009627.rn.1 TC1300001437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009628.rn.1 TC1300001437.rn.1 RTA TC
          Rat Expression Set 230 1387721_PM_at - -
          Rat Expression Set 230 1387721_at - -
          Rat Expression Set 230 1370584_a_at - -
          Rat Expression Set 230 1370584_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10767771 - -
          Rat Genome U34 Set M64299_at - -
          Rat Genome U34 Set M64299_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21248940 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367299 - -
          RatRef-12 Expression BeadChip ILMN_62348 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline