Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567882_m1
          See other AADAT GE Assays ›
          Gene Symbol:
          Aadat
          Gene Name:
          aminoadipate aminotransferase
          Gene Aliases:
          Kat2
          Chromosome Location:
          Chr.16: 32832001 - 32868680 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567882_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aadat
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017193.1 NP_058889.1 2 - 3 344 99
          XM_006253067.2 XP_006253129.1 2 - 3 432 99
          GenBank mRNA BC078864.1 - 2 - 3 340 7109210 99
          Z50144.1 - 2 - 3 344 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001529.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aadat

          Panther Classification:

          Molecular Function -

          transaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          2-oxoglutarate metabolic process
          glutamate metabolic process
          biosynthetic process
          tryptophan catabolic process to kynurenine
          L-lysine catabolic process to acetyl-CoA via saccharopine
          kynurenine metabolic process
          L-kynurenine metabolic process
          transaminase activity
          kynurenine-oxoglutarate transaminase activity
          pyridoxal phosphate binding
          protein homodimerization activity
          2-aminoadipate transaminase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P682853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266679 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177154 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89995 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13910 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P682853 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P682853 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P682853 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P991924 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700181768 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67399 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41198 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64467 - -
          Agilent Rat cDNA Microarray (G4105A) 700181768 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10791490 - -
          Affymetrix Rat Gene 1.0 ST Array 10791487 - -
          Affymetrix Rat Gene 1.0 ST Array 10791480 - -
          Affymetrix Rat Gene 1.0 ST Array 10791481 - -
          Affymetrix Rat Gene 1.0 ST Array 10791483 - -
          Affymetrix Rat Gene 1.0 ST Array 10791491 - -
          Affymetrix Rat Gene 1.0 ST Array 10791484 - -
          Affymetrix Rat Gene 1.0 ST Array 10791488 - -
          Affymetrix Rat Gene 1.0 ST Array 10791492 - -
          Affymetrix Rat Gene 1.0 ST Array 10791482 - -
          Affymetrix Rat Gene 1.0 ST Array 10791479 - -
          Affymetrix Rat Gene 1.0 ST Array 10791485 - -
          Affymetrix Rat Gene 1.0 ST Array 10791486 - -
          Affymetrix Rat Gene 1.0 ST Array 10791489 - -
          Affymetrix Rat Gene 1.1 ST Array 10791478 - -
          Rat Genome 230 2.0 Array 1387223_at - -
          Rat Genome 230 2.0 Array 1387223_PM_at - -
          Affymetrix HT Rat Focus Array 1387223_PM_at - -
          Affymetrix HT Rat Focus Array 1387223_at - -
          Affymetrix Rat Clariom S Assay PSR1600011758.rn.1 TC1600001529.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600011757.rn.1 TC1600001529.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600011761.rn.1 TC1600001529.rn.1 RTA TC
          Rat Expression Set 230 1387223_at - -
          Rat Expression Set 230 1387223_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10791478 - -
          Rat Genome U34 Set Z50144_g_at - -
          Rat Genome U34 Set Z50144_at - -
          Rat Genome U34 Set rc_AI233086_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21129651 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369433 - -
          RatRef-12 Expression BeadChip ILMN_69467 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline