Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571092_m1
          See other SLC14A1 GE Assays ›
          Gene Symbol:
          Slc14a1
          Gene Name:
          solute carrier family 14 (urea transporter), member 1
          Gene Aliases:
          HUT11, UT-B, UT3, UTB1, UrT2
          Chromosome Location:
          Chr.18: 74461064 - 74504475 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571092_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc14a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019346.2 NP_062219.2 4 - 5 585 92
          XM_008772168.1 XP_008770390.1 5 - 6 803 92
          XM_008772169.1 XP_008770391.1 4 - 5 1717 92
          XM_008772170.1 XP_008770392.1 4 - 5 654 92
          GenBank mRNA FQ211495.1 - 4 - 5 618 92
          U81518.1 - 4 - 5 555 92
          X98399.1 - 4 - 5 585 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001757.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc14a1

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          water transport
          urea transport
          transmembrane transport
          urea transmembrane transport
          water transmembrane transporter activity
          urea transmembrane transporter activity
          urea channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P340820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373401 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14676 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45549 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87534 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P340820 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P340820 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P340820 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12031 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18286 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36552 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61122 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10805338 - -
          Affymetrix Rat Gene 1.0 ST Array 10805339 - -
          Affymetrix Rat Gene 1.0 ST Array 10805342 - -
          Affymetrix Rat Gene 1.0 ST Array 10805344 - -
          Affymetrix Rat Gene 1.0 ST Array 10805336 - -
          Affymetrix Rat Gene 1.0 ST Array 10805337 - -
          Affymetrix Rat Gene 1.0 ST Array 10805340 - -
          Affymetrix Rat Gene 1.0 ST Array 10805341 - -
          Affymetrix Rat Gene 1.0 ST Array 10805343 - -
          Affymetrix Rat Gene 1.1 ST Array 10805335 - -
          Rat Genome 230 2.0 Array 1369857_PM_a_at - -
          Rat Genome 230 2.0 Array 1369857_a_at - -
          Affymetrix HT Rat Focus Array 1369857_a_at - -
          Affymetrix HT Rat Focus Array 1369857_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1800012229.rn.1 TC1800001757.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800012227.rn.1 TC1800001757.rn.1 RTA TC
          Rat Expression Set 230 1369857_PM_a_at - -
          Rat Expression Set 230 1369857_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10805335 - -
          Rat Genome U34 Set U81518_at - -
          Rat Genome U34 Set X98399cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21832307 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366471 - -
          RatRef-12 Expression BeadChip ILMN_49479 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline