Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571586_m1
          See other ACPP GE Assays ›
          Gene Symbol:
          Acpp
          Gene Name:
          acid phosphatase, prostate
          Gene Aliases:
          5'-NT, Acpp11, FRAP, Ppal, RNACPP11, pap
          Chromosome Location:
          Chr.8: 112838629 - 112884062 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571586_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acpp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134901.1 NP_001128373.1 1 - 2 122 80
          NM_020072.1 NP_064457.1 1 - 2 162 80
          GenBank mRNA DQ826426.1 - 1 - 2 122 80
          M32397.1 - 1 - 2 162 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003289.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acpp

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleobase metabolic process
          thiamine metabolic process
          nucleotide metabolic process
          dephosphorylation
          adenosine metabolic process
          regulation of sensory perception of pain
          positive regulation of adenosine receptor signaling pathway
          acid phosphatase activity
          5'-nucleotidase activity
          phosphatase activity
          choline binding
          thiamine phosphate phosphatase activity
          identical protein binding
          protein homodimerization activity
          lysophosphatidic acid phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P004458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110104 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157235 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135533 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45354 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19511 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139624 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004458 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004458 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P004458 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P332505 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34124 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5836 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60082 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9575 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10919786 - -
          Affymetrix Rat Gene 1.0 ST Array 10919779 - -
          Affymetrix Rat Gene 1.0 ST Array 10919784 - -
          Affymetrix Rat Gene 1.0 ST Array 10919781 - -
          Affymetrix Rat Gene 1.0 ST Array 10919776 - -
          Affymetrix Rat Gene 1.0 ST Array 10919787 - -
          Affymetrix Rat Gene 1.0 ST Array 10919782 - -
          Affymetrix Rat Gene 1.0 ST Array 10919785 - -
          Affymetrix Rat Gene 1.0 ST Array 10919783 - -
          Affymetrix Rat Gene 1.0 ST Array 10919778 - -
          Affymetrix Rat Gene 1.0 ST Array 10919780 - -
          Affymetrix Rat Gene 1.0 ST Array 10919777 - -
          Affymetrix Rat Gene 1.1 ST Array 10919775 - -
          Rat Genome 230 2.0 Array 1368789_PM_at - -
          Rat Genome 230 2.0 Array 1368789_at - -
          Affymetrix HT Rat Focus Array 1368789_PM_at - -
          Affymetrix HT Rat Focus Array 1368789_at - -
          Affymetrix Rat Clariom S Assay PSR0800026275.rn.1 TC0800003289.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800026273.rn.1 TC0800003289.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800026274.rn.1 TC0800003289.rn.1 RTA TC
          Rat Expression Set 230 1368789_at - -
          Rat Expression Set 230 1368789_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10919775 - -
          Rat Genome U34 Set M32397_g_at - -
          Rat Genome U34 Set M32397_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21613213 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354676 - -
          RatRef-12 Expression BeadChip ILMN_54963 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline