Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571934_m1
          See other AADAC GE Assays ›
          Gene Symbol:
          Aadac
          Gene Name:
          arylacetamide deacetylase
          Gene Aliases:
          Aada
          Chromosome Location:
          Chr.2: 150146234 - 150157480 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571934_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aadac
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020538.1 NP_065413.1 1 - 2 160 116
          GenBank mRNA AF182426.1 - 1 - 2 160 116
          BC088143.1 - 1 - 2 161 7323135 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001234.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aadac

          Panther Classification:

          Molecular Function -

          deacetylase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          metabolic process
          positive regulation of triglyceride catabolic process
          triglyceride lipase activity
          serine hydrolase activity
          deacetylase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P575398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660443 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131595 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P575398 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P575398 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P575398 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P575398 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94510 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33175 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10815627 - -
          Affymetrix Rat Gene 1.0 ST Array 10815624 - -
          Affymetrix Rat Gene 1.0 ST Array 10815625 - -
          Affymetrix Rat Gene 1.0 ST Array 10815623 - -
          Affymetrix Rat Gene 1.0 ST Array 10815626 - -
          Affymetrix Rat Gene 1.0 ST Array 10815628 - -
          Affymetrix Rat Gene 1.0 ST Array 10815629 - -
          Affymetrix Rat Gene 1.1 ST Array 10815622 - -
          Rat Genome 230 2.0 Array 1369492_PM_at - -
          Rat Genome 230 2.0 Array 1369492_at - -
          Affymetrix HT Rat Focus Array 1369492_PM_at - -
          Affymetrix HT Rat Focus Array 1369492_at - -
          Affymetrix Rat Clariom S Assay PSR0200007759.rn.1 TC0200001234.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200007762.rn.1 TC0200001234.rn.1 RTA TC
          Rat Expression Set 230 1369492_PM_at - -
          Rat Expression Set 230 1369492_at - -
          Affymetrix Rat Gene 1.0 ST Array 10815622 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22037443 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348794 - -
          RatRef-12 Expression BeadChip ILMN_58352 - -

          Back To Top
           

          Citations (1)

          Causes and prevention of tamoxifen-induced accumulation of triacylglycerol in rat liver.
          Authors
          Gudbrandsen OA, Rost TH, Berge RK
          Journal
          J Lipid Res - Page No 2223 (Sun Oct 01 00:00:00 EDT 2006)
          Volume (Year)
          47 (2006)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline