Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572267_m1
          See other POLD1 GE Assays ›
          Gene Symbol:
          Pold1
          Gene Name:
          polymerase (DNA directed), delta 1, catalytic subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 100538066 - 100554096 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572267_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pold1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021662.2 NP_067694.1 9 - 10 1233 57
          XM_006229142.2 XP_006229204.1 10 - 11 1325 57
          XM_006229143.2 XP_006229205.1 10 - 11 2842 57
          GenBank mRNA AJ222691.1 - 9 - 10 1237 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005354.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pold1

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA synthesis involved in DNA repair
          DNA catabolic process, exonucleolytic
          DNA replication
          DNA-dependent DNA replication
          base-excision repair, gap-filling
          nucleotide-excision repair, DNA gap filling
          DNA replication proofreading
          fatty acid homeostasis
          DNA biosynthetic process
          nucleic acid phosphodiester bond hydrolysis
          nucleotide binding
          DNA binding
          chromatin binding
          DNA-directed DNA polymerase activity
          3'-5'-exodeoxyribonuclease activity
          3'-5' exonuclease activity
          metal ion binding
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645704 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54277 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12090 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12090 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12090 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12090 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85874 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11890 - -
          Agilent Rat cDNA Microarray (G4105A) 700330042 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10721395 - -
          Affymetrix Rat Gene 1.0 ST Array 10721415 - -
          Affymetrix Rat Gene 1.0 ST Array 10721398 - -
          Affymetrix Rat Gene 1.0 ST Array 10721402 - -
          Affymetrix Rat Gene 1.0 ST Array 10721399 - -
          Affymetrix Rat Gene 1.0 ST Array 10721400 - -
          Affymetrix Rat Gene 1.0 ST Array 10721416 - -
          Affymetrix Rat Gene 1.0 ST Array 10721407 - -
          Affymetrix Rat Gene 1.0 ST Array 10721406 - -
          Affymetrix Rat Gene 1.0 ST Array 10721403 - -
          Affymetrix Rat Gene 1.0 ST Array 10721419 - -
          Affymetrix Rat Gene 1.0 ST Array 10721418 - -
          Affymetrix Rat Gene 1.0 ST Array 10721420 - -
          Affymetrix Rat Gene 1.0 ST Array 10721396 - -
          Affymetrix Rat Gene 1.0 ST Array 10721412 - -
          Affymetrix Rat Gene 1.0 ST Array 10721417 - -
          Affymetrix Rat Gene 1.0 ST Array 10721414 - -
          Affymetrix Rat Gene 1.0 ST Array 10721409 - -
          Affymetrix Rat Gene 1.0 ST Array 10721410 - -
          Affymetrix Rat Gene 1.0 ST Array 10721404 - -
          Affymetrix Rat Gene 1.0 ST Array 10721411 - -
          Affymetrix Rat Gene 1.0 ST Array 10721408 - -
          Affymetrix Rat Gene 1.0 ST Array 10721401 - -
          Affymetrix Rat Gene 1.0 ST Array 10721413 - -
          Affymetrix Rat Gene 1.0 ST Array 10721405 - -
          Affymetrix Rat Gene 1.0 ST Array 10721397 - -
          Affymetrix Rat Gene 1.1 ST Array 10721394 - -
          Rat Genome 230 2.0 Array 1370127_at - -
          Rat Genome 230 2.0 Array 1370127_PM_at - -
          Affymetrix HT Rat Focus Array 1370127_PM_at - -
          Affymetrix HT Rat Focus Array 1370127_at - -
          Affymetrix Rat Clariom S Assay PSR0100043210.rn.1 TC0100005354.rn.1 RTA TC
          Rat Expression Set 230 1370127_at - -
          Rat Expression Set 230 1370127_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10721394 - -
          Rat Genome U34 Set AJ222691cds_at - -
          Rat Genome U34 Set AJ222691cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20892009 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368379 - -
          RatRef-12 Expression BeadChip ILMN_49314 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline