Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572330_m1
          See other TCAM1 GE Assays ›
          Gene Symbol:
          Tcam1
          Gene Name:
          testicular cell adhesion molecule 1
          Gene Aliases:
          Tcam1p
          Chromosome Location:
          Chr.10: 94465920 - 94478591 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572330_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcam1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021673.1 NP_067705.1 2 - 3 323 68
          XM_006247642.2 XP_006247704.1 3 - 4 1077 68
          GenBank mRNA BC166495.1 - 3 - 4 385 7111123 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001789.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcam1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          single organismal cell-cell adhesion
          integrin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509277 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31854 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15545 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15545 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15545 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15545 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 559 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45757 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10739159 - -
          Affymetrix Rat Gene 1.0 ST Array 10739156 - -
          Affymetrix Rat Gene 1.0 ST Array 10739158 - -
          Affymetrix Rat Gene 1.0 ST Array 10739153 - -
          Affymetrix Rat Gene 1.0 ST Array 10739157 - -
          Affymetrix Rat Gene 1.0 ST Array 10739155 - -
          Affymetrix Rat Gene 1.0 ST Array 10739154 - -
          Affymetrix Rat Gene 1.1 ST Array 10739152 - -
          Rat Genome 230 2.0 Array 1370270_PM_at - -
          Rat Genome 230 2.0 Array 1370270_at - -
          Affymetrix HT Rat Focus Array 1370270_at - -
          Affymetrix HT Rat Focus Array 1370270_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000016387.rn.1 TC1000001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016389.rn.1 TC1000001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016382.rn.1 TC1000001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016390.rn.1 TC1000001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016379.rn.1 TC1000001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016386.rn.1 TC1000001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016381.rn.1 TC1000001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016384.rn.1 TC1000001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016385.rn.1 TC1000001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016388.rn.1 TC1000001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016383.rn.1 TC1000001789.rn.1 RTA TC
          Rat Expression Set 230 1370270_at - -
          Rat Expression Set 230 1370270_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10739152 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21069958 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362984 - -
          RatRef-12 Expression BeadChip ILMN_56257 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline