Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572371_m1
          See other EPB41L1 GE Assays ›
          Gene Symbol:
          Epb41l1
          Gene Name:
          erythrocyte membrane protein band 4.1-like 1
          Gene Aliases:
          4.1N, Epb4.1l1
          Chromosome Location:
          Chr.3: 152552814 - 152622047 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572371_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Epb41l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021681.2 NP_067713.2 24 - 25 4808 59
          NM_172090.3 NP_742087.1 21 - 22 2775 59
          XM_008762381.1 XP_008760603.1 24 - 25 4808 59
          GenBank mRNA AB019256.1 - 21 - 22 2773 59
          AB019257.1 - 24 - 25 4808 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002152.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Epb41l1

          Gene Ontology Categories:

          Function(s) Process(es)

          cortical actin cytoskeleton organization
          actin binding
          receptor binding
          structural molecule activity
          cytoskeletal protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P103289 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P305135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187313 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120888 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85720 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56394 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165655 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32021 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139727 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119871 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130188 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P103289 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P305135 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P103289 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P305135 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P305135 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P103289 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P305135 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P672622 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701333001 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24425 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5736 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67800 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18338 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71653 - -
          Agilent Rat cDNA Microarray (G4105A) 701333001 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10841557 - -
          Affymetrix Rat Gene 1.0 ST Array 10841537 - -
          Affymetrix Rat Gene 1.0 ST Array 10841551 - -
          Affymetrix Rat Gene 1.0 ST Array 10841559 - -
          Affymetrix Rat Gene 1.0 ST Array 10841533 - -
          Affymetrix Rat Gene 1.0 ST Array 10841536 - -
          Affymetrix Rat Gene 1.0 ST Array 10841538 - -
          Affymetrix Rat Gene 1.0 ST Array 10841549 - -
          Affymetrix Rat Gene 1.0 ST Array 10841535 - -
          Affymetrix Rat Gene 1.0 ST Array 10841553 - -
          Affymetrix Rat Gene 1.0 ST Array 10841546 - -
          Affymetrix Rat Gene 1.0 ST Array 10841534 - -
          Affymetrix Rat Gene 1.0 ST Array 10841548 - -
          Affymetrix Rat Gene 1.0 ST Array 10841560 - -
          Affymetrix Rat Gene 1.0 ST Array 10841541 - -
          Affymetrix Rat Gene 1.0 ST Array 10841554 - -
          Affymetrix Rat Gene 1.0 ST Array 10841539 - -
          Affymetrix Rat Gene 1.0 ST Array 10841545 - -
          Affymetrix Rat Gene 1.0 ST Array 10841556 - -
          Affymetrix Rat Gene 1.0 ST Array 10841544 - -
          Affymetrix Rat Gene 1.0 ST Array 10841550 - -
          Affymetrix Rat Gene 1.0 ST Array 10841555 - -
          Affymetrix Rat Gene 1.0 ST Array 10841542 - -
          Affymetrix Rat Gene 1.0 ST Array 10841547 - -
          Affymetrix Rat Gene 1.0 ST Array 10841558 - -
          Affymetrix Rat Gene 1.0 ST Array 10841552 - -
          Affymetrix Rat Gene 1.0 ST Array 10841543 - -
          Affymetrix Rat Gene 1.0 ST Array 10841540 - -
          Affymetrix Rat Gene 1.1 ST Array 10841532 - -
          Rat Genome 230 2.0 Array 1369212_PM_s_at - -
          Rat Genome 230 2.0 Array 1390452_PM_at - -
          Rat Genome 230 2.0 Array 1390452_at - -
          Rat Genome 230 2.0 Array 1369212_s_at - -
          Affymetrix HT Rat Focus Array 1369212_PM_s_at - -
          Affymetrix HT Rat Focus Array 1369212_s_at - -
          Affymetrix HT Rat Focus Array 1390452_at - -
          Affymetrix HT Rat Focus Array 1390452_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300015887.rn.1 TC0300002152.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300015888.rn.1 TC0300002152.rn.1 RTA TC
          Rat Expression Set 230 1369212_PM_s_at - -
          Rat Expression Set 230 1369212_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10841532 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21991968 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376582 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375856 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375679 - -
          RatRef-12 Expression BeadChip ILMN_59509 - -
          RatRef-12 Expression BeadChip ILMN_50676 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline