Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572505_m1
          See other ARHGEF1 GE Assays ›
          Gene Symbol:
          Arhgef1
          Gene Name:
          Rho guanine nucleotide exchange factor (GEF) 1
          Gene Aliases:
          Lsc
          Chromosome Location:
          Chr.1: 81769212 - 81797237 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572505_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arhgef1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021694.3 NP_067726.3 13 - 14 1374 84
          GenBank mRNA AJ236911.1 - 11 - 12 1265 84
          BC091218.1 - 13 - 14 1374 7107802 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001062.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arhgef1

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          termination of G-protein coupled receptor signaling pathway
          negative regulation of cell death
          Rho guanyl-nucleotide exchange factor activity
          GTPase activator activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P051336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753682 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90415 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56590 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172316 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051336 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051336 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P051336 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P508294 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P506852 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700528751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19697 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19836 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51645 - -
          Agilent Rat cDNA Microarray (G4105A) 700528751 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10705091 - -
          Affymetrix Rat Gene 1.0 ST Array 10705099 - -
          Affymetrix Rat Gene 1.0 ST Array 10705078 - -
          Affymetrix Rat Gene 1.0 ST Array 10705080 - -
          Affymetrix Rat Gene 1.0 ST Array 10705089 - -
          Affymetrix Rat Gene 1.0 ST Array 10705084 - -
          Affymetrix Rat Gene 1.0 ST Array 10705098 - -
          Affymetrix Rat Gene 1.0 ST Array 10705075 - -
          Affymetrix Rat Gene 1.0 ST Array 10705094 - -
          Affymetrix Rat Gene 1.0 ST Array 10705090 - -
          Affymetrix Rat Gene 1.0 ST Array 10705072 - -
          Affymetrix Rat Gene 1.0 ST Array 10705085 - -
          Affymetrix Rat Gene 1.0 ST Array 10705074 - -
          Affymetrix Rat Gene 1.0 ST Array 10705088 - -
          Affymetrix Rat Gene 1.0 ST Array 10705087 - -
          Affymetrix Rat Gene 1.0 ST Array 10705086 - -
          Affymetrix Rat Gene 1.0 ST Array 10705077 - -
          Affymetrix Rat Gene 1.0 ST Array 10705083 - -
          Affymetrix Rat Gene 1.0 ST Array 10705073 - -
          Affymetrix Rat Gene 1.0 ST Array 10705081 - -
          Affymetrix Rat Gene 1.0 ST Array 10705092 - -
          Affymetrix Rat Gene 1.0 ST Array 10705095 - -
          Affymetrix Rat Gene 1.0 ST Array 10705076 - -
          Affymetrix Rat Gene 1.0 ST Array 10705093 - -
          Affymetrix Rat Gene 1.0 ST Array 10705082 - -
          Affymetrix Rat Gene 1.0 ST Array 10705097 - -
          Affymetrix Rat Gene 1.0 ST Array 10705079 - -
          Affymetrix Rat Gene 1.0 ST Array 10705096 - -
          Affymetrix Rat Gene 1.1 ST Array 10705071 - -
          Rat Genome 230 2.0 Array 1370002_at - -
          Rat Genome 230 2.0 Array 1370002_PM_at - -
          Affymetrix HT Rat Focus Array 1370002_PM_at - -
          Affymetrix HT Rat Focus Array 1370002_at - -
          Affymetrix Rat Clariom S Assay PSR0100007581.rn.1 TC0100001062.rn.1 RTA TC
          Rat Expression Set 230 1370002_at - -
          Rat Expression Set 230 1370002_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10705071 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21724115 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376334 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline