Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573586_m1
          See other PIK3R3 GE Assays ›
          Gene Symbol:
          Pik3r3
          Gene Name:
          phosphoinositide-3-kinase, regulatory subunit 3 (gamma)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 135069965 - 135145633 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573586_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pik3r3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022213.1 NP_071549.1 9 - 10 1191 109
          XM_006238714.2 XP_006238776.1 10 - 11 1212 109
          XM_008763979.1 XP_008762201.1 10 - 11 1414 109
          GenBank mRNA D64047.1 - 9 - 10 1191 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001263.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pik3r3

          Panther Classification:

          Molecular Function -

          kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          insulin receptor signaling pathway
          phosphorylation
          regulation of phosphatidylinositol 3-kinase activity
          kinase activity
          phosphatidylinositol 3-kinase regulator activity
          1-phosphatidylinositol-3-kinase regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P343060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262066 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133916 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69307 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69709 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21677 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128745 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37361 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P343060 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P343060 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P343060 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P343060 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89934 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42486 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38119 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91769 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27774 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61799 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53497 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10871236 - -
          Affymetrix Rat Gene 1.0 ST Array 10871239 - -
          Affymetrix Rat Gene 1.0 ST Array 10871230 - -
          Affymetrix Rat Gene 1.0 ST Array 10871232 - -
          Affymetrix Rat Gene 1.0 ST Array 10871235 - -
          Affymetrix Rat Gene 1.0 ST Array 10871231 - -
          Affymetrix Rat Gene 1.0 ST Array 10871233 - -
          Affymetrix Rat Gene 1.0 ST Array 10871238 - -
          Affymetrix Rat Gene 1.0 ST Array 10871237 - -
          Affymetrix Rat Gene 1.0 ST Array 10871234 - -
          Affymetrix Rat Gene 1.1 ST Array 10871229 - -
          Rat Genome 230 2.0 Array 1369518_PM_at - -
          Rat Genome 230 2.0 Array 1369518_at - -
          Affymetrix HT Rat Focus Array 1369518_at - -
          Affymetrix HT Rat Focus Array 1369518_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500008685.rn.1 TC0500001263.rn.1 RTA TC
          Rat Expression Set 230 1369518_at - -
          Rat Expression Set 230 1369518_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10871229 - -
          Rat Genome U34 Set D64047_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20807913 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358018 - -
          RatRef-12 Expression BeadChip ILMN_50851 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline