Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573861_m1
          See other ENPEP GE Assays ›
          Gene Symbol:
          Enpep
          Gene Name:
          glutamyl aminopeptidase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 233667253 - 233743866 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573861_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Enpep
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022251.2 NP_071587.2 7 - 8 1841 73
          GenBank mRNA AF146518.1 - 7 - 8 1203 73
          AF214568.1 - 7 - 8 1658 73
          BC066663.1 - 7 - 8 1663 5623429 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004673.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Enpep

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          regulation of systemic arterial blood pressure by renin-angiotensin
          proteolysis
          cellular aromatic compound metabolic process
          cell proliferation
          cell migration
          glomerulus development
          aminopeptidase activity
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          peptide binding
          metalloaminopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P209788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39743 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P209788 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P209788 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P209788 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P144857 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P209788 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15289 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10826724 - -
          Affymetrix Rat Gene 1.1 ST Array 10826723 - -
          Rat Genome 230 2.0 Array 1368512_PM_a_at - -
          Rat Genome 230 2.0 Array 1368513_PM_at - -
          Rat Genome 230 2.0 Array 1368513_at - -
          Rat Genome 230 2.0 Array 1368512_a_at - -
          Affymetrix HT Rat Focus Array 1368513_at - -
          Affymetrix HT Rat Focus Array 1368513_PM_at - -
          Affymetrix HT Rat Focus Array 1368512_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368512_a_at - -
          Affymetrix Rat Clariom S Assay PSR0200032500.rn.1 TC0200004673.rn.1 RTA TC
          Rat Expression Set 230 1368513_PM_at - -
          Rat Expression Set 230 1368512_a_at - -
          Rat Expression Set 230 1368512_PM_a_at - -
          Rat Expression Set 230 1368513_at - -
          Affymetrix Rat Gene 1.0 ST Array 10826723 - -
          Rat Genome U34 Set S73583_at - -
          Rat Genome U34 Set rc_AI009666_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20888046 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650764 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline