Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574069_m1
          See other CASP8 GE Assays ›
          Gene Symbol:
          Casp8
          Gene Name:
          caspase 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 65614142 - 65662624 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574069_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Casp8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022277.1 NP_071613.1 5 - 6 919 86
          XM_006245014.2 XP_006245076.1 5 - 6 901 86
          XM_006245015.2 XP_006245077.1 5 - 6 1042 86
          XM_008767135.1 XP_008765357.1 5 - 6 901 86
          GenBank mRNA AF279308.1 - 5 - 6 919 86
          AF288372.1 - 5 - 6 654 86
          FQ226407.1 - 5 - 6 933 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000713.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protease Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Casp8

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          neural tube formation
          proteolysis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          heart development
          extrinsic apoptotic signaling pathway via death domain receptors
          response to cold
          macrophage differentiation
          response to cobalt ion
          response to estradiol
          response to lipopolysaccharide
          response to tumor necrosis factor
          TRAIL-activated apoptotic signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          response to ethanol
          positive regulation of macrophage differentiation
          positive regulation of proteolysis
          response to antibiotic
          protein heterooligomerization
          proteolysis involved in cellular protein catabolic process
          negative regulation of necroptotic process
          regulation of thymocyte apoptotic process
          cellular response to mechanical stimulus
          cellular response to organic cyclic compound
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          execution phase of apoptosis
          activation of cysteine-type endopeptidase activity
          hepatocyte apoptotic process
          regulation of apoptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway
          endopeptidase activity
          cysteine-type endopeptidase activity
          death receptor binding
          tumor necrosis factor receptor binding
          protein binding
          peptidase activity
          ubiquitin protein ligase binding
          protein complex binding
          death effector domain binding
          protein heterodimerization activity
          scaffold protein binding
          cysteine-type endopeptidase activity involved in apoptotic process
          cysteine-type endopeptidase activity involved in apoptotic signaling pathway

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P298210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103182 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165650 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139284 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178977 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37718 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98770 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150329 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P298210 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P298210 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P298210 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P298210 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19567 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49932 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74488 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10923605 - -
          Affymetrix Rat Gene 1.0 ST Array 10923598 - -
          Affymetrix Rat Gene 1.0 ST Array 10923597 - -
          Affymetrix Rat Gene 1.0 ST Array 10923601 - -
          Affymetrix Rat Gene 1.0 ST Array 10923596 - -
          Affymetrix Rat Gene 1.0 ST Array 10923603 - -
          Affymetrix Rat Gene 1.0 ST Array 10923602 - -
          Affymetrix Rat Gene 1.0 ST Array 10923599 - -
          Affymetrix Rat Gene 1.0 ST Array 10923604 - -
          Affymetrix Rat Gene 1.0 ST Array 10923600 - -
          Affymetrix Rat Gene 1.1 ST Array 10923595 - -
          Rat Genome 230 2.0 Array 1369262_at - -
          Rat Genome 230 2.0 Array 1369262_PM_at - -
          Affymetrix HT Rat Focus Array 1369262_at - -
          Affymetrix HT Rat Focus Array 1369262_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900005807.rn.1 TC0900000713.rn.1 RTA TC
          Rat Expression Set 230 1369262_PM_at - -
          Rat Expression Set 230 1369262_at - -
          Affymetrix Rat Gene 1.0 ST Array 10923595 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21425495 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371431 - -
          RatRef-12 Expression BeadChip ILMN_51224 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline