Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574412_m1
          See other PMPCB GE Assays ›
          Gene Symbol:
          Pmpcb
          Gene Name:
          peptidase (mitochondrial processing) beta
          Gene Aliases:
          Mppb
          Chromosome Location:
          Chr.4: 9908838 - 9921615 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574412_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pmpcb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022395.2 NP_071790.1 1 - 2 115 115
          GenBank mRNA BC078826.1 - 1 - 2 110 7113892 115
          D13907.1 - 1 - 2 93 115
          L12965.1 - 1 - 2 116 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002219.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pmpcb

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          metalloendopeptidase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690499 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90814 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12222 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12222 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12222 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12222 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700626228 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30899 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57696 - -
          Agilent Rat cDNA Microarray (G4105A) 700626228 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10860208 - -
          Affymetrix Rat Gene 1.0 ST Array 10860204 - -
          Affymetrix Rat Gene 1.0 ST Array 10860214 - -
          Affymetrix Rat Gene 1.0 ST Array 10860209 - -
          Affymetrix Rat Gene 1.0 ST Array 10860210 - -
          Affymetrix Rat Gene 1.0 ST Array 10860206 - -
          Affymetrix Rat Gene 1.0 ST Array 10860211 - -
          Affymetrix Rat Gene 1.0 ST Array 10860207 - -
          Affymetrix Rat Gene 1.0 ST Array 10860215 - -
          Affymetrix Rat Gene 1.0 ST Array 10860216 - -
          Affymetrix Rat Gene 1.0 ST Array 10860213 - -
          Affymetrix Rat Gene 1.0 ST Array 10860212 - -
          Affymetrix Rat Gene 1.1 ST Array 10860203 - -
          Rat Genome 230 2.0 Array 1383569_at - -
          Rat Genome 230 2.0 Array 1387781_at - -
          Rat Genome 230 2.0 Array 1383569_PM_at - -
          Rat Genome 230 2.0 Array 1387781_PM_at - -
          Affymetrix HT Rat Focus Array 1387781_PM_at - -
          Affymetrix HT Rat Focus Array 1383569_at - -
          Affymetrix HT Rat Focus Array 1383569_PM_at - -
          Affymetrix HT Rat Focus Array 1387781_at - -
          Affymetrix Rat Clariom S Assay PSR0400015815.rn.1 TC0400002219.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015817.rn.1 TC0400002219.rn.1 RTA TC
          Rat Expression Set 230 1387781_at - -
          Rat Expression Set 230 1387781_PM_at - -
          Rat Expression Set 230 1383569_at - -
          Rat Expression Set 230 1383569_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10860203 - -
          Rat Genome U34 Set D13907_at - -
          Rat Genome U34 Set D13907_g_at - -
          Rat Genome U34 Set rc_AI169688_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21024018 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365519 - -
          RatRef-12 Expression BeadChip ILMN_54864 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline