Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574785_m1
          See other PRDX3 GE Assays ›
          Gene Symbol:
          Prdx3
          Gene Name:
          peroxiredoxin 3
          Gene Aliases:
          Prx3
          Chromosome Location:
          Chr.1: 282238774 - 282251193 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574785_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prdx3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022540.1 NP_071985.1 5 - 6 583 90
          GenBank mRNA AF106944.1 - 5 - 6 583 90
          BC060567.1 - 5 - 6 584 6922338 90
          FQ212826.1 - 5 - 6 563 90
          FQ219655.1 - 5 - 6 580 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007992.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prdx3

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          maternal placenta development
          response to oxidative stress
          mitochondrion organization
          positive regulation of cell proliferation
          peptidyl-cysteine oxidation
          myeloid cell differentiation
          response to lipopolysaccharide
          negative regulation of kinase activity
          cellular response to oxidative stress
          cellular response to reactive oxygen species
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of NF-kappaB transcription factor activity
          regulation of mitochondrial membrane potential
          oxidation-reduction process
          molecular_function
          peroxidase activity
          protein C-terminus binding
          thioredoxin peroxidase activity
          kinase binding
          protein kinase binding
          identical protein binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          peroxiredoxin activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78879 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7773 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12243 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12243 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12243 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12243 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700302588 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81915 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58377 - -
          Agilent Rat cDNA Microarray (G4105A) 700302588 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10731212 - -
          Affymetrix Rat Gene 1.0 ST Array 10731214 - -
          Affymetrix Rat Gene 1.0 ST Array 10731211 - -
          Affymetrix Rat Gene 1.0 ST Array 10731215 - -
          Affymetrix Rat Gene 1.0 ST Array 10731216 - -
          Affymetrix Rat Gene 1.0 ST Array 10731217 - -
          Affymetrix Rat Gene 1.0 ST Array 10731219 - -
          Affymetrix Rat Gene 1.0 ST Array 10731218 - -
          Affymetrix Rat Gene 1.1 ST Array 10731210 - -
          Rat Genome 230 2.0 Array 1367591_PM_at - -
          Rat Genome 230 2.0 Array 1367591_at - -
          Affymetrix HT Rat Focus Array 1367591_PM_at - -
          Affymetrix HT Rat Focus Array 1367591_at - -
          Affymetrix Rat Clariom S Assay PSR0100064388.rn.1 TC0100007992.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100064389.rn.1 TC0100007992.rn.1 RTA TC
          Rat Expression Set 230 1367591_at - -
          Rat Expression Set 230 1367591_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10731210 - -
          Rat Genome U34 Set rc_AA799650_g_at - -
          Rat Genome U34 Set rc_AA799650_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22117960 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349059 - -
          RatRef-12 Expression BeadChip ILMN_49237 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline