Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575101_m1
          See other NDUFV3 GE Assays ›
          Gene Symbol:
          Ndufv3
          Gene Name:
          NADH dehydrogenase (ubiquinone) flavoprotein 3
          Gene Aliases:
          Mipp65, Ndufv3l
          Chromosome Location:
          Chr.20: 10265826 - 10275298 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575101_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufv3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022607.2 NP_072129.2 2 - 3 266 76
          GenBank mRNA AB000098.1 - 2 - 3 197 76
          CK365764.1 - 2 - 3 277 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000364.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndufv3

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Process(es)

          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P141541 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P535085 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P103707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257980 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173775 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141541 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P535085 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P103707 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P103707 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141541 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P535085 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P103707 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P535085 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P141541 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1007131 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700182609 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20042 - -
          Agilent Rat cDNA Microarray (G4105A) 700182609 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10829161 - -
          Affymetrix Rat Gene 1.0 ST Array 10829162 - -
          Affymetrix Rat Gene 1.0 ST Array 10829159 - -
          Affymetrix Rat Gene 1.0 ST Array 10829160 - -
          Affymetrix Rat Gene 1.0 ST Array 10767822 - -
          Affymetrix Rat Gene 1.1 ST Array 10829158 - -
          Affymetrix Rat Gene 1.1 ST Array 10767821 - -
          Rat Genome 230 2.0 Array 1368567_at - -
          Rat Genome 230 2.0 Array 1368566_a_at - -
          Rat Genome 230 2.0 Array 1368566_PM_a_at - -
          Rat Genome 230 2.0 Array 1368567_PM_at - -
          Affymetrix HT Rat Focus Array 1368566_a_at - -
          Affymetrix HT Rat Focus Array 1368567_at - -
          Affymetrix HT Rat Focus Array 1368566_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368567_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000003454.rn.1 TC2000000364.rn.1 RTA TC
          Rat Expression Set 230 1368566_a_at - -
          Rat Expression Set 230 1368566_PM_a_at - -
          Rat Expression Set 230 1368567_at - -
          Rat Expression Set 230 1368567_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10829158 - -
          Affymetrix Rat Gene 1.0 ST Array 10767821 - -
          Rat Genome U34 Set AB000098_at - -
          Rat Genome U34 Set AB000098_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22013882 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039465 - -
          RatRef-12 Expression BeadChip ILMN_58162 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline