Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575519_m1
          See other BAD GE Assays ›
          Gene Symbol:
          Bad
          Gene Name:
          BCL2-associated agonist of cell death
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 222197829 - 222207454 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575519_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bad
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022698.1 NP_073189.1 2 - 3 301 57
          XM_006230896.2 XP_006230958.1 2 - 3 1006 57
          XM_006230897.2 XP_006230959.1 1 - 2 184 57
          XM_006230898.2 XP_006230960.1 2 - 3 199 57
          GenBank mRNA AF003523.1 - 2 - 3 301 57
          AF279910.1 - 2 - 3 301 57
          AF279911.1 - 2 - 3 301 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003120.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bad

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          release of cytochrome c from mitochondria
          glucose catabolic process
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cell proliferation
          extrinsic apoptotic signaling pathway via death domain receptors
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to hormone
          response to glucose
          response to organic substance
          positive regulation of mitochondrial membrane potential
          response to organic cyclic compound
          suppression by virus of host apoptotic process
          cytokine-mediated signaling pathway
          positive regulation of insulin secretion
          response to estradiol
          response to progesterone
          positive regulation of glucokinase activity
          response to testosterone
          response to oleic acid
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          response to drug
          response to hydrogen peroxide
          glucose homeostasis
          regulation of apoptotic process
          positive regulation of apoptotic process
          response to amino acid
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          type B pancreatic cell proliferation
          response to ethanol
          positive regulation of B cell differentiation
          positive regulation of T cell differentiation
          positive regulation of proteolysis
          ADP metabolic process
          ATP metabolic process
          regulation of mitochondrial membrane permeability
          pore complex assembly
          positive regulation of epithelial cell proliferation
          response to glucocorticoid
          response to calcium ion
          positive regulation of apoptotic process by virus
          cellular process regulating host cell cycle in response to virus
          cellular response to chromate
          cellular response to mechanical stimulus
          cellular response to nicotine
          cellular response to lipid
          cellular response to hypoxia
          positive regulation of release of cytochrome c from mitochondria
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          intrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity
          positive regulation of neuron death
          positive regulation of type B pancreatic cell development
          protein binding
          phospholipid binding
          lipid binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          protein kinase binding
          protein phosphatase binding
          protein phosphatase 2B binding
          protein kinase B binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P991565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39105 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70246 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P991565 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P991565 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P991565 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P199624 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P991565 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92045 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75971 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713461 - -
          Affymetrix Rat Gene 1.0 ST Array 10713465 - -
          Affymetrix Rat Gene 1.0 ST Array 10713464 - -
          Affymetrix Rat Gene 1.0 ST Array 10713459 - -
          Affymetrix Rat Gene 1.0 ST Array 10713460 - -
          Affymetrix Rat Gene 1.0 ST Array 10713466 - -
          Affymetrix Rat Gene 1.0 ST Array 10713462 - -
          Affymetrix Rat Gene 1.0 ST Array 10713463 - -
          Affymetrix Rat Gene 1.1 ST Array 10713458 - -
          Rat Genome 230 2.0 Array 1386991_a_at - -
          Rat Genome 230 2.0 Array 1386991_PM_a_at - -
          Affymetrix HT Rat Focus Array 1386991_PM_a_at - -
          Affymetrix HT Rat Focus Array 1386991_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100025528.rn.1 TC0100003120.rn.1 RTA TC
          Rat Expression Set 230 1386991_PM_a_at - -
          Rat Expression Set 230 1386991_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713458 - -
          Rat Genome U34 Set AF003523_s_at - -
          Rat Genome U34 Set rc_AA818072_s_at - -
          Rat Genome U34 Set rc_AA818072_s_at - -
          Rat Genome U34 Set AF003523_s_at - -
          Rat Genome U34 Set AF003523_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22308589 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369751 - -
          RatRef-12 Expression BeadChip ILMN_55487 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0