Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576661_m1
          See other ARHGEF9 GE Assays ›
          Gene Symbol:
          Arhgef9
          Gene Name:
          Cdc42 guanine nucleotide exchange factor (GEF) 9
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 64249576 - 64428444 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576661_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arhgef9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023957.1 NP_076447.1 2 - 3 587 62
          GenBank mRNA AJ250425.1 - 2 - 3 587 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001882.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arhgef9

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor clustering
          Rho guanyl-nucleotide exchange factor activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P085732 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P117318 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P536115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114676 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29444 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56353 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16455 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81714 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37500 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104305 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17917 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30043 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9721 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32860 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43220 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71634 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P536115 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085732 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117318 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P536115 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117318 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085732 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P117318 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P536115 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P085732 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P536115 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702028596 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702029538 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80551 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15990 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85246 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95251 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53372 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89767 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62298 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10788 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101689 - -
          Agilent Rat cDNA Microarray (G4105A) 702028596 - -
          Agilent Rat cDNA Microarray (G4105A) 702029538 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10938470 - -
          Affymetrix Rat Gene 1.0 ST Array 10938471 - -
          Affymetrix Rat Gene 1.0 ST Array 10938468 - -
          Affymetrix Rat Gene 1.0 ST Array 10938474 - -
          Affymetrix Rat Gene 1.0 ST Array 10938475 - -
          Affymetrix Rat Gene 1.0 ST Array 10938472 - -
          Affymetrix Rat Gene 1.0 ST Array 10938476 - -
          Affymetrix Rat Gene 1.0 ST Array 10938478 - -
          Affymetrix Rat Gene 1.0 ST Array 10938469 - -
          Affymetrix Rat Gene 1.0 ST Array 10938477 - -
          Affymetrix Rat Gene 1.0 ST Array 10938473 - -
          Affymetrix Rat Gene 1.1 ST Array 10938467 - -
          Rat Genome 230 2.0 Array 1397853_PM_s_at - -
          Rat Genome 230 2.0 Array 1387199_PM_a_at - -
          Rat Genome 230 2.0 Array 1397853_s_at - -
          Rat Genome 230 2.0 Array 1387199_a_at - -
          Affymetrix HT Rat Focus Array 1397853_s_at - -
          Affymetrix HT Rat Focus Array 1387199_a_at - -
          Affymetrix HT Rat Focus Array 1387199_PM_a_at - -
          Affymetrix HT Rat Focus Array 1397853_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR0X00011935.rn.1 TC0X00001882.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00011938.rn.1 TC0X00001882.rn.1 RTA TC
          Rat Expression Set 230 1387199_a_at - -
          Rat Expression Set 230 1387199_PM_a_at - -
          Rat Expression Set 230 1397853_PM_s_at - -
          Rat Expression Set 230 1397853_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10938467 - -
          Rat Genome U34 Set rc_AI639196_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22294491 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349180 - -
          RatRef-12 Expression BeadChip ILMN_63975 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline