Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576832_m1
          See other APAF1 GE Assays ›
          Gene Symbol:
          Apaf1
          Gene Name:
          apoptotic peptidase activating factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 31699311 - 31784192 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576832_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apaf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023979.1 NP_076469.1 22 - 23 3702 95
          XM_008765263.1 XP_008763485.1 19 - 20 3494 95
          GenBank mRNA AF320222.1 - 22 - 23 3702 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002457.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apaf1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          neural tube closure
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          multicellular organism development
          brain development
          aging
          response to nutrient
          activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
          cardiac muscle cell apoptotic process
          cell differentiation
          forebrain development
          glial cell apoptotic process
          protein homooligomerization
          neuron apoptotic process
          cellular response to transforming growth factor beta stimulus
          apoptotic signaling pathway
          regulation of apoptotic DNA fragmentation
          positive regulation of apoptotic signaling pathway
          protein binding
          ATP binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          heat shock protein binding
          identical protein binding
          ADP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P125033 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141067 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157167 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16520 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30730 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52730 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1842 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50050 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165006 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12379 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125033 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12379 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125033 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P125033 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12379 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12379 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22071 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27264 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53572 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31115 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5127 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83969 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62060 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10901803 - -
          Affymetrix Rat Gene 1.0 ST Array 10901802 - -
          Affymetrix Rat Gene 1.0 ST Array 10901809 - -
          Affymetrix Rat Gene 1.0 ST Array 10901795 - -
          Affymetrix Rat Gene 1.0 ST Array 10901804 - -
          Affymetrix Rat Gene 1.0 ST Array 10901800 - -
          Affymetrix Rat Gene 1.0 ST Array 10901784 - -
          Affymetrix Rat Gene 1.0 ST Array 10901810 - -
          Affymetrix Rat Gene 1.0 ST Array 10901805 - -
          Affymetrix Rat Gene 1.0 ST Array 10901788 - -
          Affymetrix Rat Gene 1.0 ST Array 10901794 - -
          Affymetrix Rat Gene 1.0 ST Array 10901791 - -
          Affymetrix Rat Gene 1.0 ST Array 10901789 - -
          Affymetrix Rat Gene 1.0 ST Array 10901796 - -
          Affymetrix Rat Gene 1.0 ST Array 10901808 - -
          Affymetrix Rat Gene 1.0 ST Array 10901786 - -
          Affymetrix Rat Gene 1.0 ST Array 10901799 - -
          Affymetrix Rat Gene 1.0 ST Array 10901798 - -
          Affymetrix Rat Gene 1.0 ST Array 10901787 - -
          Affymetrix Rat Gene 1.0 ST Array 10901807 - -
          Affymetrix Rat Gene 1.0 ST Array 10901797 - -
          Affymetrix Rat Gene 1.0 ST Array 10901785 - -
          Affymetrix Rat Gene 1.0 ST Array 10901793 - -
          Affymetrix Rat Gene 1.0 ST Array 10901801 - -
          Affymetrix Rat Gene 1.0 ST Array 10901792 - -
          Affymetrix Rat Gene 1.0 ST Array 10901806 - -
          Affymetrix Rat Gene 1.1 ST Array 10901783 - -
          Rat Genome 230 2.0 Array 1369197_PM_at - -
          Rat Genome 230 2.0 Array 1369197_at - -
          Rat Genome 230 2.0 Array 1369198_PM_at - -
          Rat Genome 230 2.0 Array 1369198_at - -
          Affymetrix HT Rat Focus Array 1369198_PM_at - -
          Affymetrix HT Rat Focus Array 1369197_at - -
          Affymetrix HT Rat Focus Array 1369198_at - -
          Affymetrix HT Rat Focus Array 1369197_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700019630.rn.1 TC0700002457.rn.1 RTA TC
          Rat Expression Set 230 1369197_PM_at - -
          Rat Expression Set 230 1369198_at - -
          Rat Expression Set 230 1369198_PM_at - -
          Rat Expression Set 230 1369197_at - -
          Affymetrix Rat Gene 1.0 ST Array 10901783 - -
          Rat Genome U34 Set rc_AI059814_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21567957 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375211 - -
          RatRef-12 Expression BeadChip ILMN_59152 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline