Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577664_m1
          See other SLC6A11 GE Assays ›
          Gene Symbol:
          Slc6a11
          Gene Name:
          solute carrier family 6 (neurotransmitter transporter), member 11
          Gene Aliases:
          Gabt4, Gat3
          Chromosome Location:
          Chr.4: 146106386 - 146223228 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577664_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc6a11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024372.2 NP_077348.1 4 - 5 622 73
          GenBank mRNA M95738.1 - 4 - 5 625 73
          M95763.1 - 4 - 5 622 73
          S42358.1 - 4 - 5 625 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001590.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc6a11

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter transport
          brain development
          gamma-aminobutyric acid transport
          response to drug
          transmembrane transport
          neurotransmitter:sodium symporter activity
          gamma-aminobutyric acid:sodium symporter activity
          neurotransmitter binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P391345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600477 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163087 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104095 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134235 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2589 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48308 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137089 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120036 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118504 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178294 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P391345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P391345 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P391345 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P391345 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14071 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75052 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91100 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25460 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45137 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7023 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4926 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96718 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34171 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10857925 - -
          Affymetrix Rat Gene 1.0 ST Array 10857922 - -
          Affymetrix Rat Gene 1.0 ST Array 10857926 - -
          Affymetrix Rat Gene 1.0 ST Array 10857917 - -
          Affymetrix Rat Gene 1.0 ST Array 10857919 - -
          Affymetrix Rat Gene 1.0 ST Array 10857920 - -
          Affymetrix Rat Gene 1.0 ST Array 10857918 - -
          Affymetrix Rat Gene 1.0 ST Array 10857929 - -
          Affymetrix Rat Gene 1.0 ST Array 10857928 - -
          Affymetrix Rat Gene 1.0 ST Array 10857924 - -
          Affymetrix Rat Gene 1.0 ST Array 10857927 - -
          Affymetrix Rat Gene 1.0 ST Array 10857923 - -
          Affymetrix Rat Gene 1.0 ST Array 10857930 - -
          Affymetrix Rat Gene 1.0 ST Array 10857921 - -
          Affymetrix Rat Gene 1.1 ST Array 10857916 - -
          Rat Genome 230 2.0 Array 1377455_at - -
          Rat Genome 230 2.0 Array 1369715_at - -
          Rat Genome 230 2.0 Array 1376244_PM_at - -
          Rat Genome 230 2.0 Array 1376244_at - -
          Rat Genome 230 2.0 Array 1377455_PM_at - -
          Rat Genome 230 2.0 Array 1369715_PM_at - -
          Affymetrix HT Rat Focus Array 1369715_at - -
          Affymetrix HT Rat Focus Array 1369715_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400011317.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011326.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011318.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011319.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011322.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011328.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011320.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011324.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011316.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011327.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011315.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011321.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011323.rn.1 TC0400001590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011325.rn.1 TC0400001590.rn.1 RTA TC
          Rat Expression Set 230 1369715_PM_at - -
          Rat Expression Set 230 1369715_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857916 - -
          Rat Genome U34 Set S42358_s_at - -
          Rat Genome U34 Set S42358_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21008166 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355693 - -
          RatRef-12 Expression BeadChip ILMN_69451 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline