Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577966_m1
          See other ZBTB10 GE Assays ›
          Gene Symbol:
          Zbtb10
          Gene Name:
          zinc finger and BTB domain containing 10
          Gene Aliases:
          Rinzf
          Chromosome Location:
          Chr.2: 94692939 - 94730976 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577966_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024489.1 NP_077815.1 1 - 2 1454 72
          GenBank mRNA AF091457.1 - 1 - 2 1454 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003258.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zbtb10

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P219926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213628 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124348 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41411 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117661 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P219926 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P219926 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P219926 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P219926 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63344 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18564 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41735 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89451 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10822337 - -
          Affymetrix Rat Gene 1.0 ST Array 10822335 - -
          Affymetrix Rat Gene 1.0 ST Array 10822333 - -
          Affymetrix Rat Gene 1.0 ST Array 10822336 - -
          Affymetrix Rat Gene 1.0 ST Array 10822332 - -
          Affymetrix Rat Gene 1.0 ST Array 10822338 - -
          Affymetrix Rat Gene 1.0 ST Array 10822334 - -
          Affymetrix Rat Gene 1.0 ST Array 10822331 - -
          Affymetrix Rat Gene 1.0 ST Array 10822339 - -
          Affymetrix Rat Gene 1.1 ST Array 10822330 - -
          Rat Genome 230 2.0 Array 1387453_at - -
          Rat Genome 230 2.0 Array 1387453_PM_at - -
          Affymetrix HT Rat Focus Array 1387453_at - -
          Affymetrix HT Rat Focus Array 1387453_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200022539.rn.1 TC0200003258.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022540.rn.1 TC0200003258.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022538.rn.1 TC0200003258.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022541.rn.1 TC0200003258.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022537.rn.1 TC0200003258.rn.1 RTA TC
          Rat Expression Set 230 1387453_at - -
          Rat Expression Set 230 1387453_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10822330 - -
          Rat Genome U34 Set rc_AI009192_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22349217 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline