Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578177_m1
          See other SLC26A5 GE Assays ›
          Gene Symbol:
          Slc26a5
          Gene Name:
          solute carrier family 26 (anion exchanger), member 5
          Gene Aliases:
          Pres, prestin
          Chromosome Location:
          Chr.4: 9821541 - 9860571 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578177_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc26a5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030840.1 NP_110467.1 6 - 7 888 70
          GenBank mRNA AF315652.1 - 1 - 2 144 70
          AJ303372.1 - 6 - 7 888 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000153.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc26a5

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          response to ischemia
          sensory perception of sound
          metabolic process
          regulation of cell shape
          response to salicylic acid
          response to auditory stimulus
          fructose transport
          ion transmembrane transport
          negative regulation of ion transmembrane transport
          response to potassium ion
          regulation of membrane potential
          response to drug
          positive regulation of cell size
          protein tetramerization
          cochlea development
          response to thyroid hormone
          anion transmembrane transport
          response to salt
          sulfate transmembrane transport
          chloride transmembrane transport
          positive regulation of cell motility
          motor activity
          transcription factor binding
          secondary active sulfate transmembrane transporter activity
          spectrin binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P447344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51302 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13740 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13518 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159875 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172206 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P447344 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P447344 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P447344 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P447344 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27700 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53677 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4273 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10853105 - -
          Affymetrix Rat Gene 1.0 ST Array 10853108 - -
          Affymetrix Rat Gene 1.0 ST Array 10853102 - -
          Affymetrix Rat Gene 1.0 ST Array 10853098 - -
          Affymetrix Rat Gene 1.0 ST Array 10853096 - -
          Affymetrix Rat Gene 1.0 ST Array 10853101 - -
          Affymetrix Rat Gene 1.0 ST Array 10853100 - -
          Affymetrix Rat Gene 1.0 ST Array 10853093 - -
          Affymetrix Rat Gene 1.0 ST Array 10853097 - -
          Affymetrix Rat Gene 1.0 ST Array 10853104 - -
          Affymetrix Rat Gene 1.0 ST Array 10853099 - -
          Affymetrix Rat Gene 1.0 ST Array 10853103 - -
          Affymetrix Rat Gene 1.0 ST Array 10853109 - -
          Affymetrix Rat Gene 1.0 ST Array 10853106 - -
          Affymetrix Rat Gene 1.0 ST Array 10853092 - -
          Affymetrix Rat Gene 1.0 ST Array 10853107 - -
          Affymetrix Rat Gene 1.0 ST Array 10853095 - -
          Affymetrix Rat Gene 1.1 ST Array 10853091 - -
          Rat Genome 230 2.0 Array 1398291_at - -
          Rat Genome 230 2.0 Array 1398291_PM_at - -
          Affymetrix HT Rat Focus Array 1398291_PM_at - -
          Affymetrix HT Rat Focus Array 1398291_at - -
          Affymetrix Rat Clariom S Assay PSR0400001159.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001168.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001171.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001173.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001176.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001160.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001161.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001165.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001174.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001175.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001177.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001163.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001166.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001169.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001162.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001167.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001172.rn.1 TC0400000153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400001170.rn.1 TC0400000153.rn.1 RTA TC
          Rat Expression Set 230 1398291_at - -
          Rat Expression Set 230 1398291_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10853091 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21921509 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361417 - -
          RatRef-12 Expression BeadChip ILMN_51602 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline