Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580123_m1
          See other KL GE Assays ›
          Gene Symbol:
          Kl
          Gene Name:
          Klotho
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 942974 - 987206 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580123_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031336.1 NP_112626.1 4 - 5 2729 98
          GenBank mRNA AB017820.1 - 4 - 5 2729 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000017.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kl

          Panther Classification:

          Molecular Function -

          glycosidase

          Gene Ontology Categories:

          Function(s) Process(es)

          acute inflammatory response
          carbohydrate metabolic process
          energy reserve metabolic process
          aging
          insulin receptor signaling pathway
          positive regulation of bone mineralization
          calcium ion homeostasis
          positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
          beta-glucuronidase activity
          signal transducer activity
          fibroblast growth factor receptor binding
          fibroblast growth factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P041134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680019 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135746 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169746 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116204 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041134 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041134 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P041134 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12558 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28286 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14219 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89226 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10759763 - -
          Affymetrix Rat Gene 1.0 ST Array 10759766 - -
          Affymetrix Rat Gene 1.0 ST Array 10759764 - -
          Affymetrix Rat Gene 1.0 ST Array 10759767 - -
          Affymetrix Rat Gene 1.0 ST Array 10759768 - -
          Affymetrix Rat Gene 1.1 ST Array 10759762 - -
          Rat Genome 230 2.0 Array 1369361_at - -
          Rat Genome 230 2.0 Array 1369361_PM_at - -
          Affymetrix HT Rat Focus Array 1369361_PM_at - -
          Affymetrix HT Rat Focus Array 1369361_at - -
          Affymetrix Rat Clariom S Assay PSR1200000133.rn.1 TC1200000017.rn.1 RTA TC
          Rat Expression Set 230 1369361_at - -
          Rat Expression Set 230 1369361_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10759762 - -
          Rat Genome U34 Set AB017820_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20986343 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353042 - -
          RatRef-12 Expression BeadChip ILMN_55514 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline