Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580158_m1
          See other PARG GE Assays ›
          Gene Symbol:
          Parg
          Gene Name:
          poly (ADP-ribose) glycohydrolase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 8350326 - 8457999 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580158_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Parg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031339.1 NP_112629.1 8 - 9 1953 83
          XM_006252669.1 XP_006252731.1 7 - 8 1623 83
          GenBank mRNA AB019366.1 - 8 - 9 1953 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000111.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Parg

          Panther Classification:

          Molecular Function -

          glycosidase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          cellular response to DNA damage stimulus
          detection of bacterium
          positive regulation of DNA repair
          chromatin binding
          poly(ADP-ribose) glycohydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P503570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701412 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58175 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37484 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69797 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157276 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136796 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77976 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97763 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91365 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57869 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97161 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173223 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171939 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P503570 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P503570 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P503570 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P503570 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701319263 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55193 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56079 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31362 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57534 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82107 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51646 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6174 - -
          Agilent Rat cDNA Microarray (G4105A) 701319263 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10786764 - -
          Affymetrix Rat Gene 1.0 ST Array 10786756 - -
          Affymetrix Rat Gene 1.0 ST Array 10786762 - -
          Affymetrix Rat Gene 1.0 ST Array 10786765 - -
          Affymetrix Rat Gene 1.0 ST Array 10786768 - -
          Affymetrix Rat Gene 1.0 ST Array 10786766 - -
          Affymetrix Rat Gene 1.0 ST Array 10786758 - -
          Affymetrix Rat Gene 1.0 ST Array 10786763 - -
          Affymetrix Rat Gene 1.0 ST Array 10786760 - -
          Affymetrix Rat Gene 1.0 ST Array 10786759 - -
          Affymetrix Rat Gene 1.0 ST Array 10786769 - -
          Affymetrix Rat Gene 1.0 ST Array 10786767 - -
          Affymetrix Rat Gene 1.0 ST Array 10786761 - -
          Affymetrix Rat Gene 1.0 ST Array 10786773 - -
          Affymetrix Rat Gene 1.0 ST Array 10786771 - -
          Affymetrix Rat Gene 1.0 ST Array 10786770 - -
          Affymetrix Rat Gene 1.0 ST Array 10786757 - -
          Affymetrix Rat Gene 1.0 ST Array 10786772 - -
          Affymetrix Rat Gene 1.1 ST Array 10786755 - -
          Rat Genome 230 2.0 Array 1370354_PM_at - -
          Rat Genome 230 2.0 Array 1370354_at - -
          Affymetrix HT Rat Focus Array 1370354_at - -
          Affymetrix HT Rat Focus Array 1370354_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600001257.rn.1 TC1600000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600001258.rn.1 TC1600000111.rn.1 RTA TC
          Rat Expression Set 230 1370354_at - -
          Rat Expression Set 230 1370354_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10786755 - -
          Rat Genome U34 Set rc_AI012311_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21567614 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349012 - -
          RatRef-12 Expression BeadChip ILMN_59423 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline