Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580635_m1
          See other AMPD3 GE Assays ›
          Gene Symbol:
          Ampd3
          Gene Name:
          adenosine monophosphate deaminase 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 175585302 - 175630479 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580635_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ampd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031544.1 NP_113732.1 5 - 6 933 74
          XM_006230010.2 XP_006230072.1 5 - 6 1701 74
          XM_006230011.2 XP_006230073.1 5 - 6 1066 74
          XM_006230012.2 XP_006230074.1 5 - 6 1536 74
          XM_006230013.2 XP_006230075.1 5 - 6 1548 74
          XM_006230014.2 XP_006230076.1 5 - 6 1452 74
          XM_006230015.2 XP_006230077.1 5 - 6 1680 74
          XM_008759688.1 XP_008757910.1 5 - 6 943 74
          GenBank mRNA U90888.1 - 5 - 6 933 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002364.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ampd3

          Panther Classification:

          Molecular Function -

          deaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          IMP biosynthetic process
          AMP catabolic process
          IMP salvage
          ADP metabolic process
          AMP metabolic process
          ATP metabolic process
          energy homeostasis
          AMP deaminase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P610788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940857 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10060 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31746 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61967 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44996 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P610788 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P610788 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P610788 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P610788 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700143329 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11853 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18606 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10232 - -
          Agilent Rat cDNA Microarray (G4105A) 700143329 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10709894 - -
          Affymetrix Rat Gene 1.0 ST Array 10709887 - -
          Affymetrix Rat Gene 1.0 ST Array 10709891 - -
          Affymetrix Rat Gene 1.0 ST Array 10709889 - -
          Affymetrix Rat Gene 1.0 ST Array 10709884 - -
          Affymetrix Rat Gene 1.0 ST Array 10709882 - -
          Affymetrix Rat Gene 1.0 ST Array 10709892 - -
          Affymetrix Rat Gene 1.0 ST Array 10709885 - -
          Affymetrix Rat Gene 1.0 ST Array 10709893 - -
          Affymetrix Rat Gene 1.0 ST Array 10709890 - -
          Affymetrix Rat Gene 1.0 ST Array 10709886 - -
          Affymetrix Rat Gene 1.0 ST Array 10709888 - -
          Affymetrix Rat Gene 1.0 ST Array 10709881 - -
          Affymetrix Rat Gene 1.0 ST Array 10709883 - -
          Affymetrix Rat Gene 1.1 ST Array 10709880 - -
          Rat Genome 230 2.0 Array 1368342_at - -
          Rat Genome 230 2.0 Array 1368342_PM_at - -
          Affymetrix HT Rat Focus Array 1368342_at - -
          Affymetrix HT Rat Focus Array 1368342_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100017913.rn.1 TC0100002364.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100017914.rn.1 TC0100002364.rn.1 RTA TC
          Rat Expression Set 230 1368342_at - -
          Rat Expression Set 230 1368342_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10709880 - -
          Rat Genome U34 Set U90888_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21328289 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369934 - -
          RatRef-12 Expression BeadChip ILMN_67984 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline