Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581388_m1
          See other MSLN GE Assays ›
          Gene Symbol:
          Msln
          Gene Name:
          mesothelin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 15119700 - 15129129 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581388_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Msln
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031658.1 NP_113846.1 3 - 4 267 80
          XM_006246021.2 XP_006246083.1 4 - 5 218 80
          XM_006246022.2 XP_006246084.1 4 - 5 225 80
          GenBank mRNA BC070934.1 - 3 - 4 228 7098161 80
          D87351.1 - 3 - 4 267 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002437.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Msln

          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          pancreas development
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160230 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12626 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12626 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12626 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12626 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701520091 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41383 - -
          Agilent Rat cDNA Microarray (G4105A) 701520091 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741498 - -
          Affymetrix Rat Gene 1.0 ST Array 10741495 - -
          Affymetrix Rat Gene 1.0 ST Array 10741501 - -
          Affymetrix Rat Gene 1.0 ST Array 10741502 - -
          Affymetrix Rat Gene 1.0 ST Array 10741493 - -
          Affymetrix Rat Gene 1.0 ST Array 10741491 - -
          Affymetrix Rat Gene 1.0 ST Array 10741503 - -
          Affymetrix Rat Gene 1.0 ST Array 10741487 - -
          Affymetrix Rat Gene 1.0 ST Array 10741496 - -
          Affymetrix Rat Gene 1.0 ST Array 10741497 - -
          Affymetrix Rat Gene 1.0 ST Array 10741489 - -
          Affymetrix Rat Gene 1.0 ST Array 10741494 - -
          Affymetrix Rat Gene 1.0 ST Array 10741500 - -
          Affymetrix Rat Gene 1.0 ST Array 10741490 - -
          Affymetrix Rat Gene 1.0 ST Array 10741499 - -
          Affymetrix Rat Gene 1.0 ST Array 10741488 - -
          Affymetrix Rat Gene 1.1 ST Array 10741486 - -
          Rat Genome 230 2.0 Array 1368441_PM_at - -
          Rat Genome 230 2.0 Array 1368441_at - -
          Affymetrix HT Rat Focus Array 1368441_at - -
          Affymetrix HT Rat Focus Array 1368441_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000021926.rn.1 TC1000002437.rn.1 RTA TC
          Rat Expression Set 230 1368441_at - -
          Rat Expression Set 230 1368441_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741486 - -
          Rat Genome U34 Set rc_C06892_at - -
          Rat Genome U34 Set C06892_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22311132 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368737 - -
          RatRef-12 Expression BeadChip ILMN_51041 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline