Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582632_m1
          See other MYBPH GE Assays ›
          Gene Symbol:
          Mybph
          Gene Name:
          myosin binding protein H
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 51034185 - 51041903 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582632_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mybph
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031813.1 NP_114001.1 7 - 8 1130 67
          XM_006249888.2 XP_006249950.1 7 - 8 1240 67
          XM_006249889.2 XP_006249951.1 7 - 8 1239 67
          GenBank mRNA AF077338.1 - 7 - 8 1130 67
          BC061993.1 - 7 - 8 1169 5599482 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000304.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mybph

          Gene Ontology Categories:

          Function(s)

          cell adhesion

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P385066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19328 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P385066 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P385066 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P385066 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P385066 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700520329 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700637504 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4573 - -
          Agilent Rat cDNA Microarray (G4105A) 700637504 - -
          Agilent Rat cDNA Microarray (G4105A) 700520329 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10764090 - -
          Affymetrix Rat Gene 1.0 ST Array 10764081 - -
          Affymetrix Rat Gene 1.0 ST Array 10764089 - -
          Affymetrix Rat Gene 1.0 ST Array 10764086 - -
          Affymetrix Rat Gene 1.0 ST Array 10764091 - -
          Affymetrix Rat Gene 1.0 ST Array 10764085 - -
          Affymetrix Rat Gene 1.0 ST Array 10764082 - -
          Affymetrix Rat Gene 1.0 ST Array 10764087 - -
          Affymetrix Rat Gene 1.0 ST Array 10764088 - -
          Affymetrix Rat Gene 1.0 ST Array 10764083 - -
          Affymetrix Rat Gene 1.0 ST Array 10764084 - -
          Affymetrix Rat Gene 1.1 ST Array 10764079 - -
          Rat Genome 230 2.0 Array 1373478_at - -
          Rat Genome 230 2.0 Array 1368966_at - -
          Rat Genome 230 2.0 Array 1373478_PM_at - -
          Rat Genome 230 2.0 Array 1368966_PM_at - -
          Affymetrix HT Rat Focus Array 1368966_at - -
          Affymetrix HT Rat Focus Array 1368966_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300001939.rn.1 TC1300000304.rn.1 RTA TC
          Rat Expression Set 230 1368966_PM_at - -
          Rat Expression Set 230 1368966_at - -
          Affymetrix Rat Gene 1.0 ST Array 10764079 - -
          Rat Genome U34 Set AF077338_at - -
          Rat Genome U34 Set AF077338_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21936895 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354472 - -
          RatRef-12 Expression BeadChip ILMN_52760 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline