Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582657_m1
          See other RBBP7 GE Assays ›
          Gene Symbol:
          Rbbp7
          Gene Name:
          retinoblastoma binding protein 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 33648682 - 33665821 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582657_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbbp7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031816.1 NP_114004.1 2 - 3 426 74
          GenBank mRNA AF090306.1 - 2 - 3 426 74
          BC062012.1 - 2 - 3 335 5621292 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001654.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbbp7

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          DNA replication
          transcription, DNA-templated
          chromatin modification
          negative regulation of cell growth
          negative regulation of transcription, DNA-templated
          response to steroid hormone
          cellular heat acclimation
          molecular_function
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P841120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543615 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174380 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P841120 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P841120 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P841120 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P254229 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700371239 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95056 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83012 - -
          Agilent Rat cDNA Microarray (G4105A) 700371239 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937818 - -
          Affymetrix Rat Gene 1.0 ST Array 10937808 - -
          Affymetrix Rat Gene 1.0 ST Array 10937814 - -
          Affymetrix Rat Gene 1.0 ST Array 10937810 - -
          Affymetrix Rat Gene 1.0 ST Array 10937812 - -
          Affymetrix Rat Gene 1.0 ST Array 10937811 - -
          Affymetrix Rat Gene 1.0 ST Array 10937815 - -
          Affymetrix Rat Gene 1.0 ST Array 10937809 - -
          Affymetrix Rat Gene 1.0 ST Array 10937816 - -
          Affymetrix Rat Gene 1.0 ST Array 10937817 - -
          Affymetrix Rat Gene 1.0 ST Array 10937806 - -
          Affymetrix Rat Gene 1.0 ST Array 10937807 - -
          Affymetrix Rat Gene 1.0 ST Array 10937813 - -
          Affymetrix Rat Gene 1.0 ST Array 10937819 - -
          Affymetrix Rat Gene 1.1 ST Array 10937805 - -
          Rat Genome 230 2.0 Array 1398768_PM_at - -
          Rat Genome 230 2.0 Array 1398768_at - -
          Affymetrix HT Rat Focus Array 1398768_PM_at - -
          Affymetrix HT Rat Focus Array 1398768_at - -
          Affymetrix Rat Clariom S Assay PSR0X00010738.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010741.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010746.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010736.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010742.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010744.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010743.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010728.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010729.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010735.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010747.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010733.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010734.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010739.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010740.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010727.rn.1 TC0X00001654.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010731.rn.1 TC0X00001654.rn.1 RTA TC
          Rat Expression Set 230 1398768_at - -
          Rat Expression Set 230 1398768_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937805 - -
          Rat Genome U34 Set AF090306_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21862279 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359993 - -
          RatRef-12 Expression BeadChip ILMN_60122 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline