Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583531_m1
          See other LSR GE Assays ›
          Gene Symbol:
          Lsr
          Gene Name:
          lipolysis stimulated lipoprotein receptor
          Gene Aliases:
          Lisch7
          Chromosome Location:
          Chr.1: 89383515 - 89399066 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583531_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lsr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032616.1 NP_116005.1 2 - 3 661 60
          XM_006228844.2 XP_006228906.1 2 - 3 685 60
          XM_006228845.2 XP_006228907.1 2 - 3 685 60
          GenBank mRNA AF119667.1 - 2 - 3 661 60
          AF119668.1 - 2 - 3 661 60
          AF119669.1 - 2 - 3 661 60
          BC100626.1 - 2 - 3 658 7936432 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005178.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lsr

          Panther Classification:

          Molecular Function -

          defense/immunity protein immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          regulation of lipid metabolic process
          fatty acid binding
          triglyceride binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P060305 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P060303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598507 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50436 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56849 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060303 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060305 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060305 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060303 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P060303 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P060305 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P237739 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104214 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37111 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10720848 - -
          Affymetrix Rat Gene 1.0 ST Array 10720855 - -
          Affymetrix Rat Gene 1.0 ST Array 10720858 - -
          Affymetrix Rat Gene 1.0 ST Array 10720856 - -
          Affymetrix Rat Gene 1.0 ST Array 10720851 - -
          Affymetrix Rat Gene 1.0 ST Array 10720850 - -
          Affymetrix Rat Gene 1.0 ST Array 10720857 - -
          Affymetrix Rat Gene 1.0 ST Array 10720854 - -
          Affymetrix Rat Gene 1.0 ST Array 10720852 - -
          Affymetrix Rat Gene 1.0 ST Array 10720853 - -
          Affymetrix Rat Gene 1.0 ST Array 10720849 - -
          Affymetrix Rat Gene 1.1 ST Array 10720847 - -
          Rat Genome 230 2.0 Array 1368368_a_at - -
          Rat Genome 230 2.0 Array 1368368_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368368_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368368_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100041778.rn.1 TC0100005178.rn.1 RTA TC
          Rat Expression Set 230 1368368_PM_a_at - -
          Rat Expression Set 230 1368368_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10720847 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20694878 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371645 - -
          RatRef-12 Expression BeadChip ILMN_57520 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline