Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn00584304_m1
See other NRN1 GE Assays ›
Gene Symbol:
Nrn1
Gene Name:
neuritin 1
Gene Aliases:
Nrn
Chromosome Location:
Chr.17: 27925054 - 28002946 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn00584304_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Nrn1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_053346.1
NP_445798.1
246
90
GenBank mRNA
BC087582.1
-
183
7315687
90
U88958.1
-
246
90
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1700001611.rn.1
Set Membership:
Ambion siRNA available
Amplicon length between 86 and 100
Mammalian Gene Collection
Recommended Assay
Rattus norvegicus
1700
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Nrn1
Gene Ontology Categories:
Function(s)
nervous system development
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Rat Genome Microarray 4x44K v2
A_43_P12779
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_43_P12779
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_43_P12779
-
-
Agilent SurePrint G3 Rat GE 8x60K Microarray
A_43_P12779
-
-
SurePrint G3 Rat Gene Expression Microarray G4131A
A_43_P12779
-
-
Matsumoto (Agilent Rat cDNA Microarray G4105A)
702082586
-
-
Agilent Rat cDNA Microarray (G4105A)
702082586
-
-
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Gene 1.0 ST Array
10940558
-
-
Affymetrix Rat Gene 1.1 ST Array
10940558
-
-
Rat Genome 230 2.0 Array
1386969_PM_at
-
-
Rat Genome 230 2.0 Array
1386969_at
-
-
Affymetrix HT Rat Focus Array
1386969_at
-
-
Affymetrix HT Rat Focus Array
1386969_PM_at
-
-
Affymetrix Rat Clariom S Assay
PSR1700011168.rn.1
TC1700001611.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1700011167.rn.1
TC1700001611.rn.1
RTA TC
Rat Expression Set 230
1386969_PM_at
-
-
Rat Expression Set 230
1386969_at
-
-
Affymetrix Rat Gene 1.0 ST Array
10940558
-
-
Rat Genome U34 Set
U88958_at
-
-
Array Name
Probe IDs
Targets
Target Type
ABI Rat Genome Survey Microarray
22130645
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline