Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584459_m1
          See other ATOX1 GE Assays ›
          Gene Symbol:
          Atox1
          Gene Name:
          antioxidant 1 copper chaperone
          Gene Aliases:
          Atx1
          Chromosome Location:
          Chr.10: 40790850 - 40805886 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584459_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atox1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053359.2 NP_445811.1 2 - 3 179 102
          GenBank mRNA AF127137.1 - 2 - 3 115 102
          AF177671.1 - 2 - 3 194 102
          BC058458.1 - 2 - 3 203 6919265 102
          BI281905.1 - 2 - 3 102
          BM392393.1 - 2 - 3 102
          FQ220104.1 - 2 - 3 180 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004404.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atox1

          Gene Ontology Categories:

          Function(s) Process(es)

          copper ion transport
          cellular copper ion homeostasis
          response to oxidative stress
          intracellular copper ion transport
          copper ion transmembrane transport
          copper ion transmembrane transporter activity
          copper ion binding
          copper chaperone activity
          copper-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P690274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67353 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171404 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P690274 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P690274 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P690274 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P690274 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69914 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10742819 - -
          Affymetrix Rat Gene 1.0 ST Array 10742817 - -
          Affymetrix Rat Gene 1.0 ST Array 10742816 - -
          Affymetrix Rat Gene 1.0 ST Array 10742815 - -
          Affymetrix Rat Gene 1.0 ST Array 10742818 - -
          Affymetrix Rat Gene 1.0 ST Array 10742814 - -
          Affymetrix Rat Gene 1.1 ST Array 10742813 - -
          Rat Genome 230 2.0 Array 1367945_PM_at - -
          Rat Genome 230 2.0 Array 1395979_PM_at - -
          Rat Genome 230 2.0 Array 1367945_at - -
          Rat Genome 230 2.0 Array 1395979_at - -
          Affymetrix HT Rat Focus Array 1367945_at - -
          Affymetrix HT Rat Focus Array 1367945_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000041390.rn.1 TC1000004404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041389.rn.1 TC1000004404.rn.1 RTA TC
          Rat Expression Set 230 1367945_at - -
          Rat Expression Set 230 1367945_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10742813 - -
          Rat Genome U34 Set rc_AA965062_at - -
          Rat Genome U34 Set rc_AI237743_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21421858 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353644 - -
          RatRef-12 Expression BeadChip ILMN_66470 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline