Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584954_m1
          See other PICK1 GE Assays ›
          Gene Symbol:
          Pick1
          Gene Name:
          protein interacting with PRKCA 1
          Gene Aliases:
          Prkcabp
          Chromosome Location:
          Chr.7: 120465472 - 120484824 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584954_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pick1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053460.1 NP_445912.1 7 - 8 636 65
          GenBank mRNA AF327562.1 - 7 - 8 636 65
          AF373289.1 - 7 - 8 558 65
          AF542094.1 - 7 - 8 606 65
          AJ240083.2 - 7 - 8 678 65
          BC072689.1 - 8 - 9 639 7105747 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001447.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pick1

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of receptor internalization
          protein phosphorylation
          protein targeting
          protein kinase C-activating G-protein coupled receptor signaling pathway
          negative regulation of gene expression
          monoamine transport
          dopamine transport
          phosphorylation
          glial cell development
          negative regulation of Arp2/3 complex-mediated actin nucleation
          cellular response to decreased oxygen levels
          cellular response to glucose starvation
          receptor clustering
          long term synaptic depression
          negative regulation of cell death
          dendritic spine organization
          dendritic spine maintenance
          G-protein coupled receptor binding
          protein kinase C binding
          receptor binding
          protein binding
          protein C-terminus binding
          kinase activity
          ATPase activity
          enzyme binding
          protein domain specific binding
          receptor tyrosine kinase binding
          G-protein coupled glutamate receptor binding
          identical protein binding
          metal ion binding
          ephrin receptor binding
          actin filament binding
          GTPase binding
          Arp2/3 complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P601217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128516 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P601217 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P601217 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P601217 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P601217 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700505810 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2084 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77118 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 931 - -
          Agilent Rat cDNA Microarray (G4105A) 700505810 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897636 - -
          Affymetrix Rat Gene 1.0 ST Array 10897631 - -
          Affymetrix Rat Gene 1.0 ST Array 10897632 - -
          Affymetrix Rat Gene 1.0 ST Array 10897638 - -
          Affymetrix Rat Gene 1.0 ST Array 10897642 - -
          Affymetrix Rat Gene 1.0 ST Array 10897635 - -
          Affymetrix Rat Gene 1.0 ST Array 10897640 - -
          Affymetrix Rat Gene 1.0 ST Array 10897629 - -
          Affymetrix Rat Gene 1.0 ST Array 10897637 - -
          Affymetrix Rat Gene 1.0 ST Array 10897639 - -
          Affymetrix Rat Gene 1.0 ST Array 10897634 - -
          Affymetrix Rat Gene 1.0 ST Array 10897633 - -
          Affymetrix Rat Gene 1.0 ST Array 10897630 - -
          Affymetrix Rat Gene 1.0 ST Array 10897641 - -
          Affymetrix Rat Gene 1.1 ST Array 10897628 - -
          Rat Genome 230 2.0 Array 1368169_PM_at - -
          Rat Genome 230 2.0 Array 1368169_at - -
          Affymetrix HT Rat Focus Array 1368169_at - -
          Affymetrix HT Rat Focus Array 1368169_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700010849.rn.1 TC0700001447.rn.1 RTA TC
          Rat Expression Set 230 1368169_PM_at - -
          Rat Expression Set 230 1368169_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897628 - -
          Rat Genome U34 Set rc_AI233722_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21020472 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361828 - -
          RatRef-12 Expression BeadChip ILMN_60585 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline