Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585211_m1
          See other CNGA3 GE Assays ›
          Gene Symbol:
          Cnga3
          Gene Name:
          cyclic nucleotide gated channel alpha 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 43807440 - 43855878 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585211_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cnga3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053495.1 NP_445947.1 4 - 5 451 59
          XM_008767042.1 XP_008765264.1 6 - 7 975 59
          XM_008767043.1 XP_008765265.1 5 - 6 861 59
          XM_008767044.1 XP_008765266.1 5 - 6 858 59
          GenBank mRNA AB002801.1 - 4 - 5 451 59
          AJ272428.1 - 6 - 7 738 59
          AJ272429.1 - 5 - 6 624 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000489.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cnga3

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          cation transport
          phototransduction, visible light
          response to corticosteroid
          response to magnesium ion
          ion transmembrane transport
          regulation of membrane potential
          retinal cone cell development
          response to cAMP
          potassium ion transmembrane transport
          cation transmembrane transport
          signal transducer activity
          intracellular cyclic nucleotide activated cation channel activity
          intracellular cAMP activated cation channel activity
          intracellular cGMP activated cation channel activity
          voltage-gated potassium channel activity
          protein C-terminus binding
          cGMP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P005812 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13247 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178425 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12541 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131709 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157584 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005812 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005812 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157584 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157584 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P005812 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P540888 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56382 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70685 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71558 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14875 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10922649 - -
          Affymetrix Rat Gene 1.0 ST Array 10922638 - -
          Affymetrix Rat Gene 1.0 ST Array 10922648 - -
          Affymetrix Rat Gene 1.0 ST Array 10922639 - -
          Affymetrix Rat Gene 1.0 ST Array 10922646 - -
          Affymetrix Rat Gene 1.0 ST Array 10922637 - -
          Affymetrix Rat Gene 1.0 ST Array 10922647 - -
          Affymetrix Rat Gene 1.0 ST Array 10922643 - -
          Affymetrix Rat Gene 1.0 ST Array 10922641 - -
          Affymetrix Rat Gene 1.0 ST Array 10922645 - -
          Affymetrix Rat Gene 1.0 ST Array 10922644 - -
          Affymetrix Rat Gene 1.1 ST Array 10922636 - -
          Rat Genome 230 2.0 Array 1387496_PM_a_at - -
          Rat Genome 230 2.0 Array 1387496_a_at - -
          Affymetrix HT Rat Focus Array 1387496_a_at - -
          Affymetrix HT Rat Focus Array 1387496_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0900003930.rn.1 TC0900000489.rn.1 RTA TC
          Rat Expression Set 230 1387496_PM_a_at - -
          Rat Expression Set 230 1387496_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10922636 - -
          Rat Genome U34 Set AB002801_at - -
          Rat Genome U34 Set AF031943_s_at - -
          Rat Genome U34 Set AB002801_at - -
          Rat Genome U34 Set AF031943_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21587508 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358426 - -
          RatRef-12 Expression BeadChip ILMN_50241 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline