Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585303_m1
          See other PRDX4 GE Assays ›
          Gene Symbol:
          Prdx4
          Gene Name:
          peroxiredoxin 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 43495457 - 43512994 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585303_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prdx4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053512.2 NP_445964.1 2 - 3 391 86
          XM_006256951.2 XP_006257013.1 2 - 3 556 86
          GenBank mRNA AF106945.1 - 2 - 3 392 86
          BC059122.1 - 2 - 3 475 6918063 86
          FQ219258.1 - 2 - 3 439 86
          FQ220216.1 - 2 - 3 445 86
          FQ220425.1 - 2 - 3 440 86
          FQ228535.1 - 2 - 3 440 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000406.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prdx4

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          male gonad development
          4-hydroxyproline metabolic process
          protein maturation by protein folding
          extracellular matrix organization
          cell redox homeostasis
          oxidation-reduction process
          reactive oxygen species metabolic process
          negative regulation of male germ cell proliferation
          peroxidase activity
          protein homodimerization activity
          peroxiredoxin activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P376502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69856 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89026 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P376502 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P376502 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P376502 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P376502 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700515485 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33221 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54038 - -
          Agilent Rat cDNA Microarray (G4105A) 700515485 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10933707 - -
          Affymetrix Rat Gene 1.0 ST Array 10933701 - -
          Affymetrix Rat Gene 1.0 ST Array 10933703 - -
          Affymetrix Rat Gene 1.0 ST Array 10933702 - -
          Affymetrix Rat Gene 1.0 ST Array 10933704 - -
          Affymetrix Rat Gene 1.0 ST Array 10933706 - -
          Affymetrix Rat Gene 1.0 ST Array 10933705 - -
          Affymetrix Rat Gene 1.0 ST Array 10933700 - -
          Affymetrix Rat Gene 1.1 ST Array 10933699 - -
          Rat Genome 230 2.0 Array 1387891_at - -
          Rat Genome 230 2.0 Array 1387891_PM_at - -
          Affymetrix HT Rat Focus Array 1387891_PM_at - -
          Affymetrix HT Rat Focus Array 1387891_at - -
          Affymetrix Rat Clariom S Assay PSR0X00002839.rn.1 TC0X00000406.rn.1 RTA TC
          Rat Expression Set 230 1387891_PM_at - -
          Rat Expression Set 230 1387891_at - -
          Affymetrix Rat Gene 1.0 ST Array 10933699 - -
          Rat Genome U34 Set rc_AI232900_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21859625 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline