Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586013_m1
          See other ACSL5 GE Assays ›
          Gene Symbol:
          Acsl5
          Gene Name:
          acyl-CoA synthetase long-chain family member 5
          Gene Aliases:
          Acs5, Facl5
          Chromosome Location:
          Chr.1: 276240565 - 276290012 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586013_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acsl5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053607.1 NP_446059.1 2 - 3 277 111
          XM_008760551.1 XP_008758773.1 2 - 3 415 111
          GenBank mRNA AB012933.1 - 2 - 3 277 111
          BC072497.1 - 2 - 3 269 7096959 111
          FQ218149.1 - 2 - 3 332 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003844.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acsl5

          Panther Classification:

          Molecular Function -

          ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          long-chain fatty acid metabolic process
          fatty acid metabolic process
          response to nutrient
          metabolic process
          phospholipid biosynthetic process
          response to sucrose
          response to glucose
          positive regulation of plasma membrane long-chain fatty acid transport
          positive regulation of triglyceride biosynthetic process
          fatty acid transport
          positive regulation of fatty acid beta-oxidation
          cellular response to insulin stimulus
          response to cholesterol
          regulation of extrinsic apoptotic signaling pathway
          catalytic activity
          long-chain fatty acid-CoA ligase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P528844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87184 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49073 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31560 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180435 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27365 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151623 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P528844 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P528844 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P528844 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P528844 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700626158 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78954 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75796 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20497 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67363 - -
          Agilent Rat cDNA Microarray (G4105A) 700626158 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10716188 - -
          Affymetrix Rat Gene 1.0 ST Array 10716171 - -
          Affymetrix Rat Gene 1.0 ST Array 10716180 - -
          Affymetrix Rat Gene 1.0 ST Array 10716183 - -
          Affymetrix Rat Gene 1.0 ST Array 10716173 - -
          Affymetrix Rat Gene 1.0 ST Array 10716172 - -
          Affymetrix Rat Gene 1.0 ST Array 10716178 - -
          Affymetrix Rat Gene 1.0 ST Array 10716185 - -
          Affymetrix Rat Gene 1.0 ST Array 10716179 - -
          Affymetrix Rat Gene 1.0 ST Array 10716174 - -
          Affymetrix Rat Gene 1.0 ST Array 10716181 - -
          Affymetrix Rat Gene 1.0 ST Array 10716184 - -
          Affymetrix Rat Gene 1.0 ST Array 10716169 - -
          Affymetrix Rat Gene 1.0 ST Array 10716175 - -
          Affymetrix Rat Gene 1.0 ST Array 10716176 - -
          Affymetrix Rat Gene 1.0 ST Array 10716170 - -
          Affymetrix Rat Gene 1.0 ST Array 10716186 - -
          Affymetrix Rat Gene 1.0 ST Array 10716187 - -
          Affymetrix Rat Gene 1.0 ST Array 10716182 - -
          Affymetrix Rat Gene 1.1 ST Array 10716168 - -
          Rat Genome 230 2.0 Array 1397375_PM_at - -
          Rat Genome 230 2.0 Array 1386926_at - -
          Rat Genome 230 2.0 Array 1386926_PM_at - -
          Rat Genome 230 2.0 Array 1397375_at - -
          Affymetrix HT Rat Focus Array 1397375_at - -
          Affymetrix HT Rat Focus Array 1397375_PM_at - -
          Affymetrix HT Rat Focus Array 1386926_PM_at - -
          Affymetrix HT Rat Focus Array 1386926_at - -
          Affymetrix Rat Clariom S Assay PSR0100031382.rn.1 TC0100003844.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100031381.rn.1 TC0100003844.rn.1 RTA TC
          Rat Expression Set 230 1386926_PM_at - -
          Rat Expression Set 230 1386926_at - -
          Rat Expression Set 230 1397375_PM_at - -
          Rat Expression Set 230 1397375_at - -
          Affymetrix Rat Gene 1.0 ST Array 10716168 - -
          Rat Genome U34 Set AB012933_g_at - -
          Rat Genome U34 Set AB012933_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21713104 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363573 - -
          RatRef-12 Expression BeadChip ILMN_63004 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline