Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586061_s1
          See other RTN4R GE Assays ›
          Gene Symbol:
          Rtn4r
          Gene Name:
          reticulon 4 receptor
          Gene Aliases:
          NgR, NgR1, Nogor
          Chromosome Location:
          Chr.11: 86992389 - 87017112 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586061_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rtn4r
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053613.1 NP_446065.1 2 753 57
          XM_008768839.1 XP_008767061.1 2 1286 57
          GenBank mRNA AF462390.1 - 2 753 57
          AY028438.1 - 2 753 57

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rtn4r

          Gene Ontology Categories:

          Function(s) Process(es)

          axonogenesis
          negative regulation of axonogenesis
          receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110582 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152214 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152214 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152214 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12871 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60932 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10587 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54318 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73487 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755620 - -
          Affymetrix Rat Gene 1.1 ST Array 10755619 - -
          Rat Genome 230 2.0 Array 1373008_PM_x_at - -
          Rat Genome 230 2.0 Array 1369293_PM_at - -
          Rat Genome 230 2.0 Array 1388972_at - -
          Rat Genome 230 2.0 Array 1373008_x_at - -
          Rat Genome 230 2.0 Array 1369293_at - -
          Rat Genome 230 2.0 Array 1388972_PM_at - -
          Affymetrix HT Rat Focus Array 1388972_PM_at - -
          Affymetrix HT Rat Focus Array 1388972_at - -
          Affymetrix HT Rat Focus Array 1369293_at - -
          Affymetrix HT Rat Focus Array 1373008_PM_x_at - -
          Affymetrix HT Rat Focus Array 1369293_PM_at - -
          Affymetrix HT Rat Focus Array 1373008_x_at - -
          Affymetrix Rat Clariom S Assay PSR1100013110.rn.1 TC1100001991.rn.1 RTA TC
          Rat Expression Set 230 1373008_x_at - -
          Rat Expression Set 230 1369293_at - -
          Rat Expression Set 230 1373008_PM_x_at - -
          Rat Expression Set 230 1369293_PM_at - -
          Rat Expression Set 230 1388972_PM_at - -
          Rat Expression Set 230 1388972_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755619 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21739085 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376294 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline