Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587001_m1
          See other PITPNB GE Assays ›
          Gene Symbol:
          Pitpnb
          Gene Name:
          phosphatidylinositol transfer protein, beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 51279194 - 51341731 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587001_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pitpnb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053742.1 NP_446194.1 7 - 8 482 62
          XM_006249522.2 XP_006249584.1 7 - 8 526 62
          XM_006249523.2 XP_006249585.1 7 - 8 526 62
          GenBank mRNA BC061538.1 - 7 - 8 514 6888321 62
          D21132.1 - 7 - 8 482 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001841.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pitpnb

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          transport
          phospholipid transport
          lipid binding
          phosphatidylinositol transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P557474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775690 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99440 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117545 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46879 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153394 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P557474 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P557474 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P557474 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P557474 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P538531 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700544679 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701733760 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 371 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41120 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62722 - -
          Agilent Rat cDNA Microarray (G4105A) 701733760 - -
          Agilent Rat cDNA Microarray (G4105A) 700544679 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10762920 - -
          Affymetrix Rat Gene 1.0 ST Array 10762922 - -
          Affymetrix Rat Gene 1.0 ST Array 10762924 - -
          Affymetrix Rat Gene 1.0 ST Array 10762921 - -
          Affymetrix Rat Gene 1.0 ST Array 10762918 - -
          Affymetrix Rat Gene 1.0 ST Array 10762917 - -
          Affymetrix Rat Gene 1.0 ST Array 10762916 - -
          Affymetrix Rat Gene 1.0 ST Array 10762927 - -
          Affymetrix Rat Gene 1.0 ST Array 10762923 - -
          Affymetrix Rat Gene 1.0 ST Array 10762925 - -
          Affymetrix Rat Gene 1.0 ST Array 10762926 - -
          Affymetrix Rat Gene 1.0 ST Array 10762919 - -
          Affymetrix Rat Gene 1.1 ST Array 10762915 - -
          Rat Genome 230 2.0 Array 1367674_PM_at - -
          Rat Genome 230 2.0 Array 1367674_at - -
          Affymetrix HT Rat Focus Array 1367674_at - -
          Affymetrix HT Rat Focus Array 1367674_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200015709.rn.1 TC1200001841.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015707.rn.1 TC1200001841.rn.1 RTA TC
          Rat Expression Set 230 1367674_at - -
          Rat Expression Set 230 1367674_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10762915 - -
          Rat Genome U34 Set rc_AA998446_s_at - -
          Rat Genome U34 Set D21132_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22167556 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349429 - -
          RatRef-12 Expression BeadChip ILMN_61037 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline