Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587037_m1
          See other UBQLN1 GE Assays ›
          Gene Symbol:
          Ubqln1
          Gene Name:
          ubiquilin 1
          Gene Aliases:
          Da41
          Chromosome Location:
          Chr.17: 6840442 - 6878844 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587037_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ubqln1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053747.2 NP_446199.2 2 - 3 472 72
          XM_006253551.2 XP_006253613.1 2 - 3 520 72
          GenBank mRNA BC072477.1 - 2 - 3 472 7103662 72
          D87950.1 - 2 - 3 332 72
          FQ222353.1 - 2 - 3 499 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000111.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ubqln1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein ubiquitination
          response to endoplasmic reticulum stress
          cellular response to hypoxia
          regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          protein binding
          kinase binding
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401958 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19903 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12993 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37324 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27652 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12922 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12922 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12922 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1046554 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12922 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89264 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73598 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10793881 - -
          Affymetrix Rat Gene 1.0 ST Array 10793879 - -
          Affymetrix Rat Gene 1.0 ST Array 10793889 - -
          Affymetrix Rat Gene 1.0 ST Array 10793880 - -
          Affymetrix Rat Gene 1.0 ST Array 10793884 - -
          Affymetrix Rat Gene 1.0 ST Array 10793888 - -
          Affymetrix Rat Gene 1.0 ST Array 10793882 - -
          Affymetrix Rat Gene 1.0 ST Array 10793883 - -
          Affymetrix Rat Gene 1.0 ST Array 10793885 - -
          Affymetrix Rat Gene 1.0 ST Array 10793886 - -
          Affymetrix Rat Gene 1.0 ST Array 10793887 - -
          Affymetrix Rat Gene 1.0 ST Array 10793890 - -
          Affymetrix Rat Gene 1.1 ST Array 10793878 - -
          Rat Genome 230 2.0 Array 1388719_PM_at - -
          Rat Genome 230 2.0 Array 1387041_at - -
          Rat Genome 230 2.0 Array 1387041_PM_at - -
          Rat Genome 230 2.0 Array 1388719_at - -
          Affymetrix HT Rat Focus Array 1387041_PM_at - -
          Affymetrix HT Rat Focus Array 1387041_at - -
          Affymetrix Rat Clariom S Assay PSR1700000871.rn.1 TC1700000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000879.rn.1 TC1700000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000880.rn.1 TC1700000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000865.rn.1 TC1700000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000873.rn.1 TC1700000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000883.rn.1 TC1700000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000870.rn.1 TC1700000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000876.rn.1 TC1700000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000884.rn.1 TC1700000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000866.rn.1 TC1700000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000868.rn.1 TC1700000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000872.rn.1 TC1700000111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000882.rn.1 TC1700000111.rn.1 RTA TC
          Rat Expression Set 230 1387041_PM_at - -
          Rat Expression Set 230 1388719_at - -
          Rat Expression Set 230 1387041_at - -
          Rat Expression Set 230 1388719_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10793878 - -
          Rat Genome U34 Set rc_AA875206_at - -
          Rat Genome U34 Set rc_AI101492_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20791704 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351250 - -
          RatRef-12 Expression BeadChip ILMN_58264 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline