Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587242_m1
          See other SERPINI1 GE Assays ›
          Gene Symbol:
          Serpini1
          Gene Name:
          serpin peptidase inhibitor, clade I (neuroserpin), member 1
          Gene Aliases:
          PI-12, raPIT5a
          Chromosome Location:
          Chr.2: 174013378 - 174111755 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587242_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpini1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053779.1 NP_446231.1 5 - 6 994 88
          XM_008761076.1 XP_008759298.1 6 - 7 1281 88
          GenBank mRNA AF193014.1 - 5 - 6 994 88
          AF193015.1 - 5 - 6 1000 88
          BC061536.1 - 5 - 6 1008 6920827 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001401.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpini1

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          regulation of cell adhesion
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P129925 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P395374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607433 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164541 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54095 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136991 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52905 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106994 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110256 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7712 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53975 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98980 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P395374 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129925 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P395374 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129925 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P395374 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P129925 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P395374 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702142773 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17237 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64680 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45088 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1400 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1420 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64869 - -
          Agilent Rat cDNA Microarray (G4105A) 702142773 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10815964 - -
          Affymetrix Rat Gene 1.0 ST Array 10815973 - -
          Affymetrix Rat Gene 1.0 ST Array 10815966 - -
          Affymetrix Rat Gene 1.0 ST Array 10815968 - -
          Affymetrix Rat Gene 1.0 ST Array 10815970 - -
          Affymetrix Rat Gene 1.0 ST Array 10815963 - -
          Affymetrix Rat Gene 1.0 ST Array 10815967 - -
          Affymetrix Rat Gene 1.0 ST Array 10815965 - -
          Affymetrix Rat Gene 1.0 ST Array 10815972 - -
          Affymetrix Rat Gene 1.0 ST Array 10815969 - -
          Affymetrix Rat Gene 1.0 ST Array 10815971 - -
          Affymetrix Rat Gene 1.1 ST Array 10815962 - -
          Rat Genome 230 2.0 Array 1368256_PM_at - -
          Rat Genome 230 2.0 Array 1387131_PM_at - -
          Rat Genome 230 2.0 Array 1368256_at - -
          Rat Genome 230 2.0 Array 1387131_at - -
          Affymetrix HT Rat Focus Array 1368256_PM_at - -
          Affymetrix HT Rat Focus Array 1368256_at - -
          Affymetrix HT Rat Focus Array 1387131_at - -
          Affymetrix HT Rat Focus Array 1387131_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200008621.rn.1 TC0200001401.rn.1 RTA TC
          Rat Expression Set 230 1387131_PM_at - -
          Rat Expression Set 230 1368256_PM_at - -
          Rat Expression Set 230 1368256_at - -
          Rat Expression Set 230 1387131_at - -
          Affymetrix Rat Gene 1.0 ST Array 10815962 - -
          Rat Genome U34 Set rc_AI229145_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21172375 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359401 - -
          RatRef-12 Expression BeadChip ILMN_56821 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0