Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00588606_m1
          See other APBB3 GE Assays ›
          Gene Symbol:
          Apbb3
          Gene Name:
          amyloid beta (A4) precursor protein-binding, family B, member 3
          Gene Aliases:
          Fe65l2
          Chromosome Location:
          Chr.18: 29497865 - 29504995 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00588606_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apbb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053957.1 NP_446409.1 12 - 13 1502 61
          XM_006254495.1 XP_006254557.1 12 - 13 1502 61
          GenBank mRNA BC097935.1 - 9 - 10 1754 61
          Y13413.1 - 12 - 13 1502 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001874.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apbb3

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          beta-amyloid binding
          protein binding
          transcription factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P477903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608235 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34373 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P477903 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P477903 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P477903 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P477903 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13329 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803967 - -
          Affymetrix Rat Gene 1.0 ST Array 10803966 - -
          Affymetrix Rat Gene 1.0 ST Array 10803969 - -
          Affymetrix Rat Gene 1.0 ST Array 10803968 - -
          Affymetrix Rat Gene 1.0 ST Array 10803974 - -
          Affymetrix Rat Gene 1.0 ST Array 10803976 - -
          Affymetrix Rat Gene 1.0 ST Array 10803970 - -
          Affymetrix Rat Gene 1.0 ST Array 10803965 - -
          Affymetrix Rat Gene 1.0 ST Array 10803975 - -
          Affymetrix Rat Gene 1.0 ST Array 10803964 - -
          Affymetrix Rat Gene 1.0 ST Array 10803971 - -
          Affymetrix Rat Gene 1.0 ST Array 10803972 - -
          Affymetrix Rat Gene 1.0 ST Array 10803962 - -
          Affymetrix Rat Gene 1.0 ST Array 10803963 - -
          Affymetrix Rat Gene 1.0 ST Array 10803973 - -
          Affymetrix Rat Gene 1.1 ST Array 10803961 - -
          Rat Genome 230 2.0 Array 1387185_PM_at - -
          Rat Genome 230 2.0 Array 1387185_at - -
          Affymetrix HT Rat Focus Array 1387185_PM_at - -
          Affymetrix HT Rat Focus Array 1387185_at - -
          Affymetrix Rat Clariom S Assay PSR1800013132.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013143.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013144.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013148.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013127.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013133.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013140.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013141.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013146.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013147.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013131.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013125.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013142.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013124.rn.1 TC1800001874.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800013130.rn.1 TC1800001874.rn.1 RTA TC
          Rat Expression Set 230 1387185_PM_at - -
          Rat Expression Set 230 1387185_at - -
          Affymetrix Rat Gene 1.0 ST Array 10803961 - -
          Rat Genome U34 Set Y13413_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20833571 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370349 - -
          RatRef-12 Expression BeadChip ILMN_66760 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline