Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589134_m1
          See other PSMC1 GE Assays ›
          Gene Symbol:
          Psmc1
          Gene Name:
          proteasome (prosome, macropain) 26S subunit, ATPase, 1
          Gene Aliases:
          s4
          Chromosome Location:
          Chr.6: 124123283 - 124135644 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589134_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057123.1 NP_476464.1 8 - 9 1047 103
          GenBank mRNA BC063157.1 - 9 - 10 1059 6919865 103
          D50696.1 - 8 - 9 1047 103
          FQ209916.1 - 9 - 10 1052 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001343.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmc1

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein catabolic process
          ATP binding
          ATPase activity
          TBP-class protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P080109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206162 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12667 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P080109 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P080109 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P080109 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P397526 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700368885 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87412 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31690 - -
          Agilent Rat cDNA Microarray (G4105A) 700368885 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10886376 - -
          Affymetrix Rat Gene 1.0 ST Array 10886367 - -
          Affymetrix Rat Gene 1.0 ST Array 10886373 - -
          Affymetrix Rat Gene 1.0 ST Array 10886375 - -
          Affymetrix Rat Gene 1.0 ST Array 10886374 - -
          Affymetrix Rat Gene 1.0 ST Array 10886377 - -
          Affymetrix Rat Gene 1.0 ST Array 10886369 - -
          Affymetrix Rat Gene 1.0 ST Array 10886368 - -
          Affymetrix Rat Gene 1.0 ST Array 10886370 - -
          Affymetrix Rat Gene 1.0 ST Array 10886371 - -
          Affymetrix Rat Gene 1.0 ST Array 10886372 - -
          Affymetrix Rat Gene 1.1 ST Array 10886366 - -
          Rat Genome 230 2.0 Array 1398792_at - -
          Rat Genome 230 2.0 Array 1398792_PM_at - -
          Affymetrix HT Rat Focus Array 1398792_at - -
          Affymetrix HT Rat Focus Array 1398792_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600009313.rn.1 TC0600001343.rn.1 RTA TC
          Rat Expression Set 230 1398792_PM_at - -
          Rat Expression Set 230 1398792_at - -
          Affymetrix Rat Gene 1.0 ST Array 10886366 - -
          Rat Genome U34 Set D50696_at - -
          Rat Genome U34 Set rc_AI045861_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21929630 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355840 - -
          RatRef-12 Expression BeadChip ILMN_50148 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline