Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590759_g1
          See other TAS1R3 GE Assays ›
          Gene Symbol:
          Tas1r3
          Gene Name:
          taste receptor, type 1, member 3
          Gene Aliases:
          T1r3
          Chromosome Location:
          Chr.5: 173307325 - 173312950 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590759_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas1r3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130818.1 NP_570831.1 4 - 5 1495 82
          XM_008764334.1 XP_008762556.1 5 - 6 2291 82
          GenBank mRNA AF456324.1 - 4 - 5 1495 82
          AY032620.1 - 4 - 5 1495 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500004214.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Ion Channel Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas1r3

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of sweet taste
          G-protein coupled receptor signaling pathway
          sensory perception of sweet taste
          sensory perception of umami taste
          taste receptor activity
          sweet taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373880 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13137 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13137 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13137 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13137 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10882232 - -
          Affymetrix Rat Gene 1.0 ST Array 10882229 - -
          Affymetrix Rat Gene 1.0 ST Array 10882233 - -
          Affymetrix Rat Gene 1.0 ST Array 10882230 - -
          Affymetrix Rat Gene 1.0 ST Array 10882228 - -
          Affymetrix Rat Gene 1.0 ST Array 10882231 - -
          Affymetrix Rat Gene 1.1 ST Array 10882227 - -
          Rat Genome 230 2.0 Array 1369397_at - -
          Rat Genome 230 2.0 Array 1369397_PM_at - -
          Affymetrix HT Rat Focus Array 1369397_at - -
          Affymetrix HT Rat Focus Array 1369397_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500032193.rn.1 TC0500004214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500032196.rn.1 TC0500004214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500032191.rn.1 TC0500004214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500032194.rn.1 TC0500004214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500032195.rn.1 TC0500004214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500032192.rn.1 TC0500004214.rn.1 RTA TC
          Rat Expression Set 230 1369397_PM_at - -
          Rat Expression Set 230 1369397_at - -
          Affymetrix Rat Gene 1.0 ST Array 10882227 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20737194 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371907 - -
          RatRef-12 Expression BeadChip ILMN_55979 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline