Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590960_m1
          See other XPNPEP1 GE Assays ›
          Gene Symbol:
          Xpnpep1
          Gene Name:
          X-prolyl aminopeptidase (aminopeptidase P) 1, soluble
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 273708217 - 273758247 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590960_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xpnpep1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_131913.1 NP_571988.1 2 - 3 161 69
          GenBank mRNA AF038591.1 - 2 - 3 161 69
          BC061758.1 - 3 - 4 271 5600093 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007893.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Xpnpep1

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          bradykinin catabolic process
          aminopeptidase activity
          manganese ion binding
          protein homodimerization activity
          metalloaminopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P562625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576678 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91143 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71442 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140377 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105385 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91399 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58307 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P562625 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P562625 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P562625 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P562625 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P251908 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701031137 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51440 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79058 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10794 - -
          Agilent Rat cDNA Microarray (G4105A) 701031137 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10730849 - -
          Affymetrix Rat Gene 1.0 ST Array 10730852 - -
          Affymetrix Rat Gene 1.0 ST Array 10730846 - -
          Affymetrix Rat Gene 1.0 ST Array 10730843 - -
          Affymetrix Rat Gene 1.0 ST Array 10730853 - -
          Affymetrix Rat Gene 1.0 ST Array 10730840 - -
          Affymetrix Rat Gene 1.0 ST Array 10730844 - -
          Affymetrix Rat Gene 1.0 ST Array 10730842 - -
          Affymetrix Rat Gene 1.0 ST Array 10730836 - -
          Affymetrix Rat Gene 1.0 ST Array 10730841 - -
          Affymetrix Rat Gene 1.0 ST Array 10730838 - -
          Affymetrix Rat Gene 1.0 ST Array 10730847 - -
          Affymetrix Rat Gene 1.0 ST Array 10730848 - -
          Affymetrix Rat Gene 1.0 ST Array 10730835 - -
          Affymetrix Rat Gene 1.0 ST Array 10730834 - -
          Affymetrix Rat Gene 1.0 ST Array 10730845 - -
          Affymetrix Rat Gene 1.0 ST Array 10730851 - -
          Affymetrix Rat Gene 1.0 ST Array 10730837 - -
          Affymetrix Rat Gene 1.0 ST Array 10730854 - -
          Affymetrix Rat Gene 1.0 ST Array 10730839 - -
          Affymetrix Rat Gene 1.0 ST Array 10730850 - -
          Affymetrix Rat Gene 1.1 ST Array 10730833 - -
          Rat Genome 230 2.0 Array 1370160_PM_at - -
          Rat Genome 230 2.0 Array 1370160_at - -
          Affymetrix HT Rat Focus Array 1370160_PM_at - -
          Affymetrix HT Rat Focus Array 1370160_at - -
          Affymetrix Rat Clariom S Assay PSR0100063696.rn.1 TC0100007893.rn.1 RTA TC
          Rat Expression Set 230 1370160_PM_at - -
          Rat Expression Set 230 1370160_at - -
          Affymetrix Rat Gene 1.0 ST Array 10730833 - -
          Rat Genome U34 Set AF038591_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21665425 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350525 - -
          RatRef-12 Expression BeadChip ILMN_49165 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline